Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137174771-137174996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC2
Entrez Gene ID
29882
Gene Name
anaphase promoting complex subunit 2
Gene Aliases
APC2
UniGene
Hs.533262
Gene Chromosome Location
Chr.9: 137174784-137188605 on Build GRCh38
Transcripts
NM_013366.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4152191 c.2427C>A p.V809V Chr.9: 137174984-137174984 on Build GRCh38
7355407 c.2461T>A p.C821S Chr.9: 137174950-137174950 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779320567 Chr.9: 137174912-137174913 on Build GRCh38
rs931096037 Chr.9: 137174974-137174974 on Build GRCh38
rs1023390225 Chr.9: 137174908-137174908 on Build GRCh38
rs1034801307 Chr.9: 137174888-137174888 on Build GRCh38
rs978779108 Chr.9: 137174881-137174881 on Build GRCh38
rs1010303859 Chr.9: 137174865-137174865 on Build GRCh38
rs993067183 Chr.9: 137174892-137174892 on Build GRCh38
rs776118220 Chr.9: 137174914-137174914 on Build GRCh38
rs367818808 Chr.9: 137174913-137174916 on Build GRCh38
rs544312391 Chr.9: 137174978-137174978 on Build GRCh38
rs922437070 Chr.9: 137174936-137174936 on Build GRCh38
rs375760748 Chr.9: 137174943-137174943 on Build GRCh38
rs747050984 Chr.9: 137174935-137174936 on Build GRCh38
rs754888219 Chr.9: 137174928-137174928 on Build GRCh38
rs975178382 Chr.9: 137174935-137174935 on Build GRCh38
rs1006836492 Chr.9: 137174824-137174824 on Build GRCh38
rs746061402 Chr.9: 137174949-137174949 on Build GRCh38
rs770260478 Chr.9: 137174963-137174963 on Build GRCh38
rs981847480 Chr.9: 137174910-137174910 on Build GRCh38
rs780285803 Chr.9: 137174972-137174972 on Build GRCh38
rs201930610 Chr.9: 137174923-137174923 on Build GRCh38
rs767321067 Chr.9: 137174922-137174922 on Build GRCh38
rs965220496 Chr.9: 137174864-137174864 on Build GRCh38
rs761356351 Chr.9: 137174921-137174921 on Build GRCh38
rs761815092 Chr.9: 137174911-137174914 on Build GRCh38
rs879238752 Chr.9: 137174831-137174831 on Build GRCh38
rs770028631 Chr.9: 137174893-137174893 on Build GRCh38
rs1020372761 Chr.9: 137174869-137174869 on Build GRCh38
rs753606898 Chr.9: 137174925-137174925 on Build GRCh38
rs970502193 Chr.9: 137174909-137174909 on Build GRCh38
rs749608183 Chr.9: 137174975-137174975 on Build GRCh38
rs1047091338 Chr.9: 137174861-137174861 on Build GRCh38
rs778777115 Chr.9: 137174931-137174931 on Build GRCh38
rs558528606 Chr.9: 137174924-137174924 on Build GRCh38
rs958965629 Chr.9: 137174889-137174889 on Build GRCh38
rs774010604 Chr.9: 137174920-137174920 on Build GRCh38
rs1013054152 Chr.9: 137174896-137174896 on Build GRCh38
rs901233054 Chr.9: 137174822-137174822 on Build GRCh38
rs763364223 Chr.9: 137174916-137174916 on Build GRCh38
rs534265577 Chr.9: 137174920-137174921 on Build GRCh38
rs1019446700 Chr.9: 137174860-137174860 on Build GRCh38
rs969090928 Chr.9: 137174880-137174880 on Build GRCh38
rs914702683 Chr.9: 137174927-137174927 on Build GRCh38
rs575091986 Chr.9: 137174956-137174956 on Build GRCh38
rs538304767 Chr.9: 137174816-137174816 on Build GRCh38
rs757293837 Chr.9: 137174941-137174941 on Build GRCh38
rs764594059 Chr.9: 137174919-137174919 on Build GRCh38
rs531945705 Chr.9: 137174917-137174920 on Build GRCh38
rs767355203 Chr.9: 137174911-137174922 on Build GRCh38
rs778001160 Chr.9: 137174913-137174928 on Build GRCh38
rs532700018 Chr.9: 137174913-137174920 on Build GRCh38
rs751672304 Chr.9: 137174932-137174932 on Build GRCh38
rs940880850 Chr.9: 137174945-137174945 on Build GRCh38
rs758632699 Chr.9: 137174913-137174924 on Build GRCh38

View Full Product Details