Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 34930968-34931193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT6B
Entrez Gene ID
10693
Gene Name
chaperonin containing TCP1 subunit 6B
Gene Aliases
CCT-zeta-2, CCTZ-2, Cctz2, TCP-1-zeta-2, TSA303
UniGene
Hs.73072
Gene Chromosome Location
Chr.17: 34927859-34981221 on Build GRCh38
Transcripts
NM_001193529.2 NM_001193530.1 NM_006584.3 XM_011524203.2 XM_017024023.1 XM_017024024.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
977754 c.1414G>A p.E472K Chr.17: 34930985-34930985 on Build GRCh38
5937713 c.1361A>T p.N454I Chr.17: 34931038-34931038 on Build GRCh38
3819294 c.1359G>A p.Q453Q Chr.17: 34931040-34931040 on Build GRCh38
4887140 c.1408C>T p.H470Y Chr.17: 34930991-34930991 on Build GRCh38
1710143 c.1351C>T p.L451F Chr.17: 34931048-34931048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368580642 Chr.17: 34931071-34931071 on Build GRCh38
rs759920774 Chr.17: 34931055-34931055 on Build GRCh38
rs537445931 Chr.17: 34931080-34931080 on Build GRCh38
rs1051672558 Chr.17: 34931079-34931080 on Build GRCh38
rs975056490 Chr.17: 34931018-34931018 on Build GRCh38
rs898149368 Chr.17: 34931081-34931081 on Build GRCh38
rs1038676502 Chr.17: 34931036-34931036 on Build GRCh38
rs557436680 Chr.17: 34931118-34931118 on Build GRCh38
rs143249349 Chr.17: 34930992-34930992 on Build GRCh38
rs763577520 Chr.17: 34931076-34931076 on Build GRCh38
rs750460352 Chr.17: 34931046-34931046 on Build GRCh38
rs777387893 Chr.17: 34931015-34931015 on Build GRCh38
rs751701658 Chr.17: 34931086-34931086 on Build GRCh38
rs770287468 Chr.17: 34931061-34931061 on Build GRCh38
rs995466231 Chr.17: 34931147-34931147 on Build GRCh38
rs780193250 Chr.17: 34931087-34931088 on Build GRCh38
rs147082978 Chr.17: 34931010-34931010 on Build GRCh38
rs1028651338 Chr.17: 34931157-34931157 on Build GRCh38
rs761940269 Chr.17: 34931100-34931100 on Build GRCh38
rs770681410 Chr.17: 34931051-34931051 on Build GRCh38
rs997027655 Chr.17: 34931077-34931077 on Build GRCh38
rs113709527 Chr.17: 34931102-34931102 on Build GRCh38
rs111371395 Chr.17: 34931043-34931043 on Build GRCh38
rs777121956 Chr.17: 34931052-34931052 on Build GRCh38
rs534850160 Chr.17: 34931004-34931004 on Build GRCh38
rs765270328 Chr.17: 34931003-34931004 on Build GRCh38
rs746609126 Chr.17: 34931050-34931050 on Build GRCh38
rs956407480 Chr.17: 34931126-34931126 on Build GRCh38
rs758640326 Chr.17: 34931080-34931081 on Build GRCh38
rs1003883226 Chr.17: 34931074-34931074 on Build GRCh38

View Full Product Details