Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17181633-17181894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CECR1
Entrez Gene ID
51816
Gene Name
cat eye syndrome chromosome region, candidate 1
Gene Aliases
ADA2, ADGF, IDGFL, PAN, SNEDS
UniGene
Hs.170310
Gene Chromosome Location
Chr.22: 17178790-17221854 on Build GRCh38
Transcripts
NM_001282225.1 NM_001282226.1 NM_001282227.1 NM_001282228.1 NM_001282229.1 NM_177405.2 XM_006724080.3 XM_011546133.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1414759 c.713C>A p.S238Y Chr.22: 17181826-17181826 on Build GRCh38
6048538 c.1392delG p.M465fs*1 Chr.22: 17181870-17181870 on Build GRCh38
1414760 c.1403A>G p.D468G Chr.22: 17181859-17181859 on Build GRCh38
4881369 c.677C>A p.A226D Chr.22: 17181862-17181862 on Build GRCh38
6048539 c.669delG p.M224fs*1 Chr.22: 17181870-17181870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369431884 Chr.22: 17181777-17181777 on Build GRCh38
rs767335027 Chr.22: 17181874-17181874 on Build GRCh38
rs757028671 Chr.22: 17181870-17181870 on Build GRCh38
rs1018298547 Chr.22: 17181841-17181841 on Build GRCh38
rs569141813 Chr.22: 17181689-17181689 on Build GRCh38
rs754904956 Chr.22: 17181859-17181865 on Build GRCh38
rs774572672 Chr.22: 17181793-17181793 on Build GRCh38
rs369910191 Chr.22: 17181832-17181832 on Build GRCh38
rs41282461 Chr.22: 17181789-17181789 on Build GRCh38
rs558205738 Chr.22: 17181806-17181806 on Build GRCh38
rs377574121 Chr.22: 17181831-17181831 on Build GRCh38
rs572691117 Chr.22: 17181774-17181774 on Build GRCh38
rs748583065 Chr.22: 17181796-17181796 on Build GRCh38
rs774380287 Chr.22: 17181729-17181729 on Build GRCh38
rs11703884 Chr.22: 17181701-17181701 on Build GRCh38
rs1012870769 Chr.22: 17181720-17181720 on Build GRCh38
rs746116155 Chr.22: 17181731-17181731 on Build GRCh38
rs372585593 Chr.22: 17181824-17181824 on Build GRCh38
rs140149635 Chr.22: 17181732-17181732 on Build GRCh38
rs369050563 Chr.22: 17181814-17181814 on Build GRCh38
rs769257505 Chr.22: 17181776-17181776 on Build GRCh38
rs138076444 Chr.22: 17181839-17181839 on Build GRCh38
rs768415914 Chr.22: 17181791-17181791 on Build GRCh38
rs962016488 Chr.22: 17181767-17181767 on Build GRCh38
rs553791075 Chr.22: 17181775-17181775 on Build GRCh38
rs376562964 Chr.22: 17181690-17181690 on Build GRCh38
rs1014574948 Chr.22: 17181754-17181754 on Build GRCh38
rs765537418 Chr.22: 17181858-17181858 on Build GRCh38
rs1003307615 Chr.22: 17181736-17181736 on Build GRCh38
rs58754958 Chr.22: 17181809-17181809 on Build GRCh38
rs200262142 Chr.22: 17181840-17181840 on Build GRCh38
rs751292513 Chr.22: 17181868-17181868 on Build GRCh38

View Full Product Details