Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 18659429-18659698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL5B
Entrez Gene ID
221079
Gene Name
ADP ribosylation factor like GTPase 5B
Gene Aliases
ARL8
UniGene
Hs.25362
Gene Chromosome Location
Chr.10: 18659335-18681639 on Build GRCh38
Transcripts
NM_178815.3 XM_005252400.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5341216 c.31C>T p.L11F Chr.10: 18659668-18659668 on Build GRCh38
5036041 c.46+1G>T p.? Chr.10: 18659684-18659684 on Build GRCh38
6065538 c.38G>T p.C13F Chr.10: 18659675-18659675 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs12767957 Chr.10: 18659653-18659653 on Build GRCh38
rs1033895732 Chr.10: 18659618-18659618 on Build GRCh38
rs778721717 Chr.10: 18659629-18659629 on Build GRCh38
rs966076309 Chr.10: 18659451-18659451 on Build GRCh38
rs774487793 Chr.10: 18659614-18659614 on Build GRCh38
rs199726601 Chr.10: 18659652-18659652 on Build GRCh38
rs768930512 Chr.10: 18659610-18659610 on Build GRCh38
rs928967709 Chr.10: 18659580-18659580 on Build GRCh38
rs754755684 Chr.10: 18659628-18659628 on Build GRCh38
rs558056174 Chr.10: 18659576-18659576 on Build GRCh38
rs984857616 Chr.10: 18659482-18659482 on Build GRCh38
rs941864368 Chr.10: 18659568-18659568 on Build GRCh38
rs761290580 Chr.10: 18659624-18659624 on Build GRCh38
rs867463251 Chr.10: 18659495-18659495 on Build GRCh38
rs889300778 Chr.10: 18659559-18659559 on Build GRCh38
rs372321983 Chr.10: 18659460-18659460 on Build GRCh38
rs543052596 Chr.10: 18659623-18659623 on Build GRCh38
rs993601906 Chr.10: 18659454-18659454 on Build GRCh38
rs556589727 Chr.10: 18659463-18659463 on Build GRCh38
rs781733930 Chr.10: 18659597-18659597 on Build GRCh38
rs1021465068 Chr.10: 18659500-18659500 on Build GRCh38
rs917786446 Chr.10: 18659503-18659503 on Build GRCh38
rs910347814 Chr.10: 18659550-18659550 on Build GRCh38
rs775884724 Chr.10: 18659605-18659605 on Build GRCh38
rs777193117 Chr.10: 18659649-18659649 on Build GRCh38
rs1008102110 Chr.10: 18659570-18659570 on Build GRCh38
rs546240282 Chr.10: 18659452-18659452 on Build GRCh38
rs538289611 Chr.10: 18659452-18659452 on Build GRCh38
rs926474521 Chr.10: 18659484-18659484 on Build GRCh38
rs571021339 Chr.10: 18659476-18659476 on Build GRCh38
rs897607640 Chr.10: 18659578-18659578 on Build GRCh38
rs142861535 Chr.10: 18659522-18659522 on Build GRCh38
rs866481286 Chr.10: 18659505-18659505 on Build GRCh38
rs976875294 Chr.10: 18659672-18659672 on Build GRCh38
rs568549564 Chr.10: 18659475-18659475 on Build GRCh38
rs767442886 Chr.10: 18659622-18659622 on Build GRCh38
rs376801401 Chr.10: 18659627-18659627 on Build GRCh38
rs144696662 Chr.10: 18659661-18659661 on Build GRCh38
rs986064529 Chr.10: 18659545-18659545 on Build GRCh38
rs770265970 Chr.10: 18659670-18659670 on Build GRCh38
rs759425966 Chr.10: 18659541-18659541 on Build GRCh38
rs893941099 Chr.10: 18659621-18659621 on Build GRCh38
rs936447437 Chr.10: 18659527-18659527 on Build GRCh38
rs545969621 Chr.10: 18659565-18659565 on Build GRCh38
rs754725063 Chr.10: 18659591-18659591 on Build GRCh38
rs190486132 Chr.10: 18659508-18659508 on Build GRCh38
rs778705410 Chr.10: 18659592-18659592 on Build GRCh38
rs535531169 Chr.10: 18659486-18659486 on Build GRCh38
rs906818595 Chr.10: 18659467-18659467 on Build GRCh38
rs770307553 Chr.10: 18659603-18659603 on Build GRCh38
rs989541134 Chr.10: 18659497-18659497 on Build GRCh38
rs972442019 Chr.10: 18659462-18659462 on Build GRCh38
rs1002316754 Chr.10: 18659468-18659468 on Build GRCh38
rs771426980 Chr.10: 18659595-18659595 on Build GRCh38
rs752349902 Chr.10: 18659636-18659636 on Build GRCh38
rs576323204 Chr.10: 18659616-18659616 on Build GRCh38
rs952015089 Chr.10: 18659594-18659594 on Build GRCh38
rs368255520 Chr.10: 18659573-18659573 on Build GRCh38
rs757961621 Chr.10: 18659643-18659643 on Build GRCh38
rs760531850 Chr.10: 18659611-18659612 on Build GRCh38

View Full Product Details