Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 100892415-100892682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH1A3
Entrez Gene ID
220
Gene Name
aldehyde dehydrogenase 1 family member A3
Gene Aliases
ALDH1A6, ALDH6, MCOP8, RALDH3
UniGene
Hs.459538
Gene Chromosome Location
Chr.15: 100879692-100916626 on Build GRCh38
Transcripts
NM_000693.3 NM_001293815.1 NR_135827.1

Gene Symbol
LOC101927751
Entrez Gene ID
101927751
Gene Name
uncharacterized LOC101927751
Gene Aliases
-
UniGene
Hs.659224
Gene Chromosome Location
Chr.15: 100892343-100919283 on Build GRCh38
Transcripts
NM_000693.3 NM_001293815.1 NR_135827.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
959587 c.377T>G p.F126C Chr.15: 100892541-100892541 on Build GRCh38
6320417 c.410G>T p.C137F Chr.15: 100892574-100892574 on Build GRCh38
1375677 c.475G>T p.D159Y Chr.15: 100892639-100892639 on Build GRCh38
5522068 c.396C>T p.I132I Chr.15: 100892560-100892560 on Build GRCh38
3401585 c.436G>T p.G146W Chr.15: 100892600-100892600 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750971909 Chr.15: 100892572-100892572 on Build GRCh38
rs78931658 Chr.15: 100892640-100892640 on Build GRCh38
rs765072555 Chr.15: 100892606-100892606 on Build GRCh38
rs769874094 Chr.15: 100892496-100892496 on Build GRCh38
rs76890635 Chr.15: 100892449-100892449 on Build GRCh38
rs754619607 Chr.15: 100892598-100892598 on Build GRCh38
rs3809521 Chr.15: 100892476-100892476 on Build GRCh38
rs142560469 Chr.15: 100892558-100892558 on Build GRCh38
rs779541443 Chr.15: 100892467-100892467 on Build GRCh38
rs918956199 Chr.15: 100892611-100892611 on Build GRCh38
rs745902602 Chr.15: 100892487-100892487 on Build GRCh38
rs201030697 Chr.15: 100892495-100892495 on Build GRCh38
rs200146843 Chr.15: 100892477-100892477 on Build GRCh38
rs760135875 Chr.15: 100892566-100892566 on Build GRCh38
rs113686214 Chr.15: 100892630-100892630 on Build GRCh38
rs753449630 Chr.15: 100892474-100892474 on Build GRCh38
rs200486213 Chr.15: 100892595-100892595 on Build GRCh38
rs867514037 Chr.15: 100892524-100892524 on Build GRCh38
rs139132913 Chr.15: 100892551-100892551 on Build GRCh38
rs769150775 Chr.15: 100892540-100892540 on Build GRCh38
rs781379161 Chr.15: 100892483-100892483 on Build GRCh38
rs772819081 Chr.15: 100892561-100892561 on Build GRCh38
rs529459760 Chr.15: 100892661-100892661 on Build GRCh38
rs758397848 Chr.15: 100892617-100892617 on Build GRCh38
rs746912683 Chr.15: 100892645-100892645 on Build GRCh38
rs201931276 Chr.15: 100892628-100892628 on Build GRCh38
rs986905739 Chr.15: 100892497-100892497 on Build GRCh38
rs756165454 Chr.15: 100892648-100892648 on Build GRCh38
rs1021397683 Chr.15: 100892437-100892437 on Build GRCh38
rs775873830 Chr.15: 100892525-100892525 on Build GRCh38
rs766385223 Chr.15: 100892596-100892598 on Build GRCh38
rs763917270 Chr.15: 100892593-100892593 on Build GRCh38
rs748148109 Chr.15: 100892482-100892482 on Build GRCh38
rs765816810 Chr.15: 100892583-100892583 on Build GRCh38
rs373337351 Chr.15: 100892567-100892567 on Build GRCh38
rs767890865 Chr.15: 100892560-100892560 on Build GRCh38
rs749466605 Chr.15: 100892530-100892530 on Build GRCh38

View Full Product Details