Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 72347971-72348206 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEXA
Entrez Gene ID
3073
Gene Name
hexosaminidase subunit alpha
Gene Aliases
TSD
UniGene
Hs.604479
Gene Chromosome Location
Chr.15: 72343435-72376473 on Build GRCh38
Transcripts
NM_000520.5 NM_001318825.1 NR_134869.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5662167 c.1034delG p.G345fs*37 Chr.15: 72348087-72348087 on Build GRCh38
77842 c.1025A>G p.K342R Chr.15: 72348096-72348096 on Build GRCh38
3370247 c.1073+1G>A p.? Chr.15: 72348047-72348047 on Build GRCh38
471028 c.1073C>T p.T358M Chr.15: 72348048-72348048 on Build GRCh38
6455463 c.1033G>A p.G345S Chr.15: 72348088-72348088 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs983475113 Chr.15: 72348156-72348156 on Build GRCh38
rs745555933 Chr.15: 72348082-72348082 on Build GRCh38
rs76173977 Chr.15: 72348047-72348047 on Build GRCh38
rs545676277 Chr.15: 72347997-72347997 on Build GRCh38
rs750575614 Chr.15: 72348132-72348132 on Build GRCh38
rs779456035 Chr.15: 72348052-72348052 on Build GRCh38
rs767713828 Chr.15: 72348120-72348120 on Build GRCh38
rs191330716 Chr.15: 72348093-72348093 on Build GRCh38
rs780502991 Chr.15: 72348044-72348044 on Build GRCh38
rs754219266 Chr.15: 72348149-72348149 on Build GRCh38
rs747036890 Chr.15: 72348026-72348026 on Build GRCh38
rs574497871 Chr.15: 72348079-72348079 on Build GRCh38
rs149121118 Chr.15: 72348107-72348107 on Build GRCh38
rs775322979 Chr.15: 72348088-72348088 on Build GRCh38
rs1055244053 Chr.15: 72348164-72348164 on Build GRCh38
rs766562662 Chr.15: 72348148-72348148 on Build GRCh38
rs1057516755 Chr.15: 72348075-72348078 on Build GRCh38
rs552505619 Chr.15: 72348048-72348048 on Build GRCh38
rs377083597 Chr.15: 72348161-72348161 on Build GRCh38
rs760923149 Chr.15: 72348142-72348142 on Build GRCh38
rs762704131 Chr.15: 72348089-72348089 on Build GRCh38
rs367719010 Chr.15: 72348007-72348007 on Build GRCh38
rs757467632 Chr.15: 72348000-72348000 on Build GRCh38
rs767271183 Chr.15: 72348010-72348010 on Build GRCh38
rs374977824 Chr.15: 72348152-72348152 on Build GRCh38
rs770433168 Chr.15: 72348075-72348075 on Build GRCh38
rs121907967 Chr.15: 72348134-72348134 on Build GRCh38
rs756371964 Chr.15: 72348030-72348030 on Build GRCh38
rs751665673 Chr.15: 72347998-72347998 on Build GRCh38
rs752508323 Chr.15: 72348095-72348096 on Build GRCh38
rs746518979 Chr.15: 72348074-72348074 on Build GRCh38
rs750658255 Chr.15: 72348024-72348024 on Build GRCh38

View Full Product Details