Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53989312-53989557 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXC10
Entrez Gene ID
3226
Gene Name
homeobox C10
Gene Aliases
HOX3I
UniGene
Hs.44276
Gene Chromosome Location
Chr.12: 53985162-53990279 on Build GRCh38
Transcripts
NM_017409.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5040265 c.905G>A p.S302N Chr.12: 53989322-53989322 on Build GRCh38
694043 c.1004T>C p.L335P Chr.12: 53989421-53989421 on Build GRCh38
3812390 c.978G>A p.M326I Chr.12: 53989395-53989395 on Build GRCh38
4575558 c.994A>G p.I332V Chr.12: 53989411-53989411 on Build GRCh38
4567853 c.897_898GG>AA p.E300K Chr.12: 53989314-53989315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375042242 Chr.12: 53989424-53989424 on Build GRCh38
rs754678586 Chr.12: 53989460-53989460 on Build GRCh38
rs781690894 Chr.12: 53989417-53989417 on Build GRCh38
rs761825384 Chr.12: 53989373-53989373 on Build GRCh38
rs780124260 Chr.12: 53989492-53989492 on Build GRCh38
rs778387407 Chr.12: 53989466-53989466 on Build GRCh38
rs747284003 Chr.12: 53989415-53989415 on Build GRCh38
rs200363226 Chr.12: 53989519-53989520 on Build GRCh38
rs775547283 Chr.12: 53989427-53989427 on Build GRCh38
rs184950567 Chr.12: 53989473-53989473 on Build GRCh38
rs141068564 Chr.12: 53989436-53989436 on Build GRCh38
rs538290710 Chr.12: 53989400-53989400 on Build GRCh38
rs373631729 Chr.12: 53989457-53989457 on Build GRCh38
rs751249692 Chr.12: 53989363-53989363 on Build GRCh38
rs760685171 Chr.12: 53989454-53989454 on Build GRCh38
rs770222472 Chr.12: 53989491-53989491 on Build GRCh38
rs764018812 Chr.12: 53989339-53989339 on Build GRCh38
rs749984625 Chr.12: 53989388-53989388 on Build GRCh38
rs767310213 Chr.12: 53989379-53989379 on Build GRCh38
rs377534424 Chr.12: 53989414-53989414 on Build GRCh38
rs761476104 Chr.12: 53989451-53989451 on Build GRCh38
rs566280089 Chr.12: 53989462-53989462 on Build GRCh38
rs765992428 Chr.12: 53989455-53989455 on Build GRCh38
rs749279055 Chr.12: 53989494-53989494 on Build GRCh38
rs752359227 Chr.12: 53989468-53989468 on Build GRCh38
rs781527084 Chr.12: 53989477-53989477 on Build GRCh38
rs763243890 Chr.12: 53989430-53989430 on Build GRCh38
rs753612424 Chr.12: 53989403-53989403 on Build GRCh38
rs1036745713 Chr.12: 53989519-53989519 on Build GRCh38
rs554485781 Chr.12: 53989521-53989521 on Build GRCh38
rs577934558 Chr.12: 53989452-53989452 on Build GRCh38
rs926127834 Chr.12: 53989481-53989481 on Build GRCh38
rs771497248 Chr.12: 53989416-53989416 on Build GRCh38
rs774219729 Chr.12: 53989449-53989449 on Build GRCh38
rs750436134 Chr.12: 53989453-53989453 on Build GRCh38
rs370667773 Chr.12: 53989463-53989463 on Build GRCh38
rs764149921 Chr.12: 53989339-53989342 on Build GRCh38
rs369495069 Chr.12: 53989426-53989426 on Build GRCh38
rs939691188 Chr.12: 53989505-53989505 on Build GRCh38
rs746445222 Chr.12: 53989489-53989489 on Build GRCh38
rs138069369 Chr.12: 53989398-53989398 on Build GRCh38
rs1048812575 Chr.12: 53989490-53989490 on Build GRCh38
rs35616568 Chr.12: 53989509-53989509 on Build GRCh38
rs754518171 Chr.12: 53989404-53989404 on Build GRCh38
rs967672222 Chr.12: 53989474-53989474 on Build GRCh38

View Full Product Details