Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74530842-74531115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTRA2
Entrez Gene ID
27429
Gene Name
HtrA serine peptidase 2
Gene Aliases
OMI, PARK13, PRSS25
UniGene
Hs.469045
Gene Chromosome Location
Chr.2: 74529405-74533556 on Build GRCh38
Transcripts
NM_001321727.1 NM_001321728.1 NM_013247.4 NM_145074.2 NR_135769.1 NR_135770.1 NR_135771.1 NR_135772.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5802973 c.803A>G p.N268S Chr.2: 74531002-74531002 on Build GRCh38
5823963 c.740G>T p.R247L Chr.2: 74530939-74530939 on Build GRCh38
722481 c.835C>G p.Q279E Chr.2: 74531034-74531034 on Build GRCh38
6449641 c.838C>G p.R280G Chr.2: 74531037-74531037 on Build GRCh38
1210141 c.782G>A p.G261E Chr.2: 74530981-74530981 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749313302 Chr.2: 74530896-74530896 on Build GRCh38
rs371440746 Chr.2: 74531051-74531051 on Build GRCh38
rs760267720 Chr.2: 74530873-74530873 on Build GRCh38
rs755898457 Chr.2: 74530988-74530988 on Build GRCh38
rs201875183 Chr.2: 74531066-74531066 on Build GRCh38
rs763567914 Chr.2: 74530938-74530938 on Build GRCh38
rs750129356 Chr.2: 74530976-74530976 on Build GRCh38
rs777650346 Chr.2: 74531024-74531024 on Build GRCh38
rs754963751 Chr.2: 74531006-74531006 on Build GRCh38
rs943608853 Chr.2: 74530920-74530920 on Build GRCh38
rs775948550 Chr.2: 74531072-74531072 on Build GRCh38
rs749157271 Chr.2: 74530998-74530998 on Build GRCh38
rs771190615 Chr.2: 74530906-74530906 on Build GRCh38
rs777079325 Chr.2: 74530915-74530915 on Build GRCh38
rs770265005 Chr.2: 74531070-74531070 on Build GRCh38
rs374733787 Chr.2: 74531062-74531062 on Build GRCh38
rs990689419 Chr.2: 74530945-74530945 on Build GRCh38
rs369105449 Chr.2: 74530925-74530925 on Build GRCh38
rs1057519082 Chr.2: 74530927-74530929 on Build GRCh38
rs573555457 Chr.2: 74530866-74530866 on Build GRCh38
rs71640292 Chr.2: 74530877-74530877 on Build GRCh38
rs763466238 Chr.2: 74530944-74530944 on Build GRCh38
rs371835841 Chr.2: 74530871-74530871 on Build GRCh38
rs142306682 Chr.2: 74530947-74530947 on Build GRCh38
rs11538692 Chr.2: 74531040-74531040 on Build GRCh38
rs773388863 Chr.2: 74530901-74530901 on Build GRCh38
rs763331417 Chr.2: 74531084-74531084 on Build GRCh38
rs747148949 Chr.2: 74530902-74530902 on Build GRCh38
rs772013612 Chr.2: 74530897-74530897 on Build GRCh38
rs779949168 Chr.2: 74530894-74530894 on Build GRCh38
rs767167271 Chr.2: 74530955-74530955 on Build GRCh38
rs759184920 Chr.2: 74530954-74530954 on Build GRCh38
rs771194907 Chr.2: 74531063-74531063 on Build GRCh38
rs995273947 Chr.2: 74530939-74530939 on Build GRCh38
rs776113129 Chr.2: 74530932-74530932 on Build GRCh38

View Full Product Details