Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130079583-130079822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR107
Entrez Gene ID
57720
Gene Name
G protein-coupled receptor 107
Gene Aliases
GCDRP, LUSTR1, bA138E2.2
UniGene
Hs.512461
Gene Chromosome Location
Chr.9: 130053706-130140169 on Build GRCh38
Transcripts
NM_001136557.1 NM_001136558.1 NM_001287346.1 NM_020960.4 XM_017014953.1 XM_017014954.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7332371 c.451G>A p.E151K Chr.9: 130079694-130079694 on Build GRCh38
4138857 c.482A>C p.N161T Chr.9: 130079725-130079725 on Build GRCh38
1208572 c.443G>A p.S148N Chr.9: 130079686-130079686 on Build GRCh38
3413379 c.514C>G p.Q172E Chr.9: 130079757-130079757 on Build GRCh38
6182792 c.522A>T p.T174T Chr.9: 130079765-130079765 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147298143 Chr.9: 130079663-130079663 on Build GRCh38
rs368852782 Chr.9: 130079748-130079748 on Build GRCh38
rs144814090 Chr.9: 130079758-130079758 on Build GRCh38
rs548172287 Chr.9: 130079720-130079720 on Build GRCh38
rs779349140 Chr.9: 130079687-130079687 on Build GRCh38
rs773995699 Chr.9: 130079766-130079766 on Build GRCh38
rs773295334 Chr.9: 130079700-130079700 on Build GRCh38
rs144029325 Chr.9: 130079637-130079637 on Build GRCh38
rs1000476900 Chr.9: 130079691-130079691 on Build GRCh38
rs912379316 Chr.9: 130079639-130079639 on Build GRCh38
rs374633031 Chr.9: 130079615-130079615 on Build GRCh38
rs367761671 Chr.9: 130079782-130079782 on Build GRCh38
rs746427159 Chr.9: 130079791-130079791 on Build GRCh38
rs776982703 Chr.9: 130079743-130079743 on Build GRCh38
rs903431312 Chr.9: 130079650-130079650 on Build GRCh38
rs559753298 Chr.9: 130079667-130079667 on Build GRCh38
rs868302696 Chr.9: 130079704-130079704 on Build GRCh38
rs144933240 Chr.9: 130079714-130079714 on Build GRCh38
rs750338919 Chr.9: 130079616-130079616 on Build GRCh38
rs772294783 Chr.9: 130079696-130079696 on Build GRCh38
rs528524605 Chr.9: 130079690-130079690 on Build GRCh38
rs756027571 Chr.9: 130079625-130079625 on Build GRCh38
rs749131292 Chr.9: 130079656-130079656 on Build GRCh38
rs751199654 Chr.9: 130079606-130079606 on Build GRCh38
rs761395806 Chr.9: 130079776-130079776 on Build GRCh38
rs747504394 Chr.9: 130079734-130079734 on Build GRCh38
rs765581698 Chr.9: 130079763-130079763 on Build GRCh38
rs756825656 Chr.9: 130079608-130079608 on Build GRCh38
rs773605805 Chr.9: 130079777-130079778 on Build GRCh38
rs1041928200 Chr.9: 130079642-130079642 on Build GRCh38
rs766954570 Chr.9: 130079777-130079777 on Build GRCh38
rs199544380 Chr.9: 130079779-130079779 on Build GRCh38
rs749905573 Chr.9: 130079778-130079778 on Build GRCh38
rs568241532 Chr.9: 130079735-130079735 on Build GRCh38
rs141062301 Chr.9: 130079666-130079666 on Build GRCh38

View Full Product Details