Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 96833004-96833251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HS6ST3
Entrez Gene ID
266722
Gene Name
heparan sulfate 6-O-sulfotransferase 3
Gene Aliases
HS6ST-3
UniGene
Hs.171001
Gene Chromosome Location
Chr.13: 96090634-96839562 on Build GRCh38
Transcripts
NM_153456.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
949054 c.1255T>C p.Y419H Chr.13: 96833037-96833037 on Build GRCh38
3955784 c.1241T>C p.L414P Chr.13: 96833023-96833023 on Build GRCh38
6487844 c.1253G>T p.R418L Chr.13: 96833035-96833035 on Build GRCh38
4691247 c.1292G>A p.R431H Chr.13: 96833074-96833074 on Build GRCh38
6336691 c.1257C>G p.Y419* Chr.13: 96833039-96833039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369533483 Chr.13: 96833087-96833087 on Build GRCh38
rs918645785 Chr.13: 96833221-96833221 on Build GRCh38
rs200967296 Chr.13: 96833157-96833157 on Build GRCh38
rs774812153 Chr.13: 96833180-96833180 on Build GRCh38
rs778905293 Chr.13: 96833086-96833086 on Build GRCh38
rs745756353 Chr.13: 96833205-96833205 on Build GRCh38
rs372819266 Chr.13: 96833102-96833102 on Build GRCh38
rs748286220 Chr.13: 96833143-96833143 on Build GRCh38
rs776338767 Chr.13: 96833169-96833169 on Build GRCh38
rs746745930 Chr.13: 96833165-96833165 on Build GRCh38
rs372992635 Chr.13: 96833109-96833109 on Build GRCh38
rs140922305 Chr.13: 96833041-96833041 on Build GRCh38
rs954259083 Chr.13: 96833126-96833126 on Build GRCh38
rs769971018 Chr.13: 96833089-96833089 on Build GRCh38
rs150223784 Chr.13: 96833093-96833093 on Build GRCh38
rs866414186 Chr.13: 96833085-96833085 on Build GRCh38
rs756354506 Chr.13: 96833075-96833075 on Build GRCh38
rs778912839 Chr.13: 96833140-96833140 on Build GRCh38
rs368666736 Chr.13: 96833225-96833225 on Build GRCh38
rs762458089 Chr.13: 96833118-96833118 on Build GRCh38
rs778619413 Chr.13: 96833074-96833074 on Build GRCh38
rs746839989 Chr.13: 96833095-96833095 on Build GRCh38
rs748257864 Chr.13: 96833045-96833045 on Build GRCh38
rs374919031 Chr.13: 96833073-96833073 on Build GRCh38
rs761117322 Chr.13: 96833224-96833224 on Build GRCh38
rs764600252 Chr.13: 96833040-96833040 on Build GRCh38
rs200033142 Chr.13: 96833106-96833106 on Build GRCh38
rs758970533 Chr.13: 96833032-96833032 on Build GRCh38
rs772945372 Chr.13: 96833094-96833094 on Build GRCh38
rs770768259 Chr.13: 96833166-96833166 on Build GRCh38
rs757636802 Chr.13: 96833055-96833055 on Build GRCh38
rs955722946 Chr.13: 96833089-96833089 on Build GRCh38
rs756562993 Chr.13: 96833130-96833130 on Build GRCh38
rs1001462839 Chr.13: 96833217-96833217 on Build GRCh38
rs573695399 Chr.13: 96833090-96833090 on Build GRCh38
rs773915233 Chr.13: 96833203-96833203 on Build GRCh38
rs891884481 Chr.13: 96833160-96833161 on Build GRCh38
rs769325134 Chr.13: 96833170-96833170 on Build GRCh38
rs773427647 Chr.13: 96833212-96833212 on Build GRCh38
rs562169523 Chr.13: 96833107-96833107 on Build GRCh38
rs200597659 Chr.13: 96833082-96833082 on Build GRCh38
rs907243781 Chr.13: 96833191-96833191 on Build GRCh38
rs774890970 Chr.13: 96833117-96833117 on Build GRCh38
rs754076410 Chr.13: 96833139-96833139 on Build GRCh38
rs758295331 Chr.13: 96833145-96833145 on Build GRCh38
rs202239358 Chr.13: 96833122-96833122 on Build GRCh38
rs768159623 Chr.13: 96833119-96833119 on Build GRCh38
rs112912651 Chr.13: 96833133-96833133 on Build GRCh38
rs946154865 Chr.13: 96833182-96833182 on Build GRCh38
rs772162176 Chr.13: 96833215-96833215 on Build GRCh38
rs1021841575 Chr.13: 96833112-96833112 on Build GRCh38
rs772651064 Chr.13: 96833186-96833186 on Build GRCh38
rs115169080 Chr.13: 96833183-96833183 on Build GRCh38
rs755071265 Chr.13: 96833083-96833083 on Build GRCh38

View Full Product Details