Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 119431474-119431719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRK5
Entrez Gene ID
2869
Gene Name
G protein-coupled receptor kinase 5
Gene Aliases
GPRK5
UniGene
Hs.524625
Gene Chromosome Location
Chr.10: 119207685-119455619 on Build GRCh38
Transcripts
NM_005308.2 XM_005269707.1 XM_005269708.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5522802 c.687C>T p.S229S Chr.10: 119431476-119431476 on Build GRCh38
4883929 c.721G>T p.V241F Chr.10: 119431510-119431510 on Build GRCh38
3806561 c.715G>A p.E239K Chr.10: 119431504-119431504 on Build GRCh38
4965672 c.738+9G>A p.? Chr.10: 119431536-119431536 on Build GRCh38
5650103 c.716_718delAGA p.K240delK Chr.10: 119431505-119431507 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561429004 Chr.10: 119431599-119431599 on Build GRCh38
rs74643082 Chr.10: 119431696-119431696 on Build GRCh38
rs754375992 Chr.10: 119431518-119431518 on Build GRCh38
rs1006238306 Chr.10: 119431598-119431598 on Build GRCh38
rs375035540 Chr.10: 119431600-119431600 on Build GRCh38
rs140741672 Chr.10: 119431538-119431538 on Build GRCh38
rs550115144 Chr.10: 119431613-119431613 on Build GRCh38
rs1022906437 Chr.10: 119431592-119431592 on Build GRCh38
rs563597805 Chr.10: 119431530-119431530 on Build GRCh38
rs777360386 Chr.10: 119431581-119431584 on Build GRCh38
rs986047396 Chr.10: 119431536-119431536 on Build GRCh38
rs533488194 Chr.10: 119431615-119431615 on Build GRCh38
rs760749742 Chr.10: 119431675-119431675 on Build GRCh38
rs7099478 Chr.10: 119431674-119431674 on Build GRCh38
rs370118216 Chr.10: 119431555-119431555 on Build GRCh38
rs1024395392 Chr.10: 119431618-119431618 on Build GRCh38
rs546977938 Chr.10: 119431626-119431626 on Build GRCh38
rs12721545 Chr.10: 119431614-119431614 on Build GRCh38
rs12721550 Chr.10: 119431561-119431561 on Build GRCh38
rs202120111 Chr.10: 119431510-119431510 on Build GRCh38
rs757441015 Chr.10: 119431559-119431559 on Build GRCh38
rs939328357 Chr.10: 119431619-119431619 on Build GRCh38
rs751637124 Chr.10: 119431556-119431556 on Build GRCh38
rs767270791 Chr.10: 119431501-119431501 on Build GRCh38
rs1051275057 Chr.10: 119431564-119431564 on Build GRCh38
rs766824009 Chr.10: 119431504-119431504 on Build GRCh38
rs140965782 Chr.10: 119431502-119431502 on Build GRCh38
rs780068014 Chr.10: 119431560-119431560 on Build GRCh38
rs948647145 Chr.10: 119431677-119431677 on Build GRCh38
rs548292082 Chr.10: 119431591-119431591 on Build GRCh38
rs778571488 Chr.10: 119431573-119431573 on Build GRCh38
rs760228524 Chr.10: 119431503-119431503 on Build GRCh38
rs567082719 Chr.10: 119431656-119431656 on Build GRCh38
rs112871819 Chr.10: 119431511-119431511 on Build GRCh38
rs990851959 Chr.10: 119431654-119431654 on Build GRCh38
rs1004387219 Chr.10: 119431519-119431519 on Build GRCh38
rs748020504 Chr.10: 119431577-119431577 on Build GRCh38
rs200647050 Chr.10: 119431554-119431554 on Build GRCh38

View Full Product Details