Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 168930622-168930895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB11
Entrez Gene ID
8647
Gene Name
ATP binding cassette subfamily B member 11
Gene Aliases
ABC16, BRIC2, BSEP, PFIC-2, PFIC2, PGY4, SPGP
UniGene
Hs.658439
Gene Chromosome Location
Chr.2: 168915468-169031396 on Build GRCh38
Transcripts
NM_003742.2 XM_006712817.3 XM_011512077.2 XM_011512078.2 XM_011512081.2 XM_017005165.1 XM_017005166.1 XM_017005167.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4828029 c.3289A>T p.N1097Y Chr.2: 168930787-168930787 on Build GRCh38
3314667 c.3321G>C p.Q1107H Chr.2: 168930755-168930755 on Build GRCh38
6494753 c.3330G>A p.A1110A Chr.2: 168930746-168930746 on Build GRCh38
5901110 c.3214-5C>T p.? Chr.2: 168930867-168930867 on Build GRCh38
3314670 c.3234T>C p.I1078I Chr.2: 168930842-168930842 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051851643 Chr.2: 168930659-168930659 on Build GRCh38
rs779530257 Chr.2: 168930705-168930705 on Build GRCh38
rs1007406465 Chr.2: 168930695-168930695 on Build GRCh38
rs762789280 Chr.2: 168930858-168930858 on Build GRCh38
rs779375126 Chr.2: 168930742-168930742 on Build GRCh38
rs1016688353 Chr.2: 168930821-168930821 on Build GRCh38
rs759634677 Chr.2: 168930668-168930668 on Build GRCh38
rs762255602 Chr.2: 168930814-168930814 on Build GRCh38
rs1016053414 Chr.2: 168930711-168930711 on Build GRCh38
rs767636463 Chr.2: 168930776-168930776 on Build GRCh38
rs953658667 Chr.2: 168930799-168930799 on Build GRCh38
rs188996270 Chr.2: 168930655-168930655 on Build GRCh38
rs373956718 Chr.2: 168930747-168930747 on Build GRCh38
rs113099601 Chr.2: 168930746-168930746 on Build GRCh38
rs757912819 Chr.2: 168930728-168930728 on Build GRCh38
rs904619956 Chr.2: 168930645-168930645 on Build GRCh38
rs368393726 Chr.2: 168930797-168930797 on Build GRCh38
rs371859686 Chr.2: 168930800-168930800 on Build GRCh38
rs1025708273 Chr.2: 168930754-168930754 on Build GRCh38
rs752974260 Chr.2: 168930669-168930669 on Build GRCh38
rs201287126 Chr.2: 168930656-168930656 on Build GRCh38
rs973967505 Chr.2: 168930832-168930832 on Build GRCh38
rs151195688 Chr.2: 168930651-168930651 on Build GRCh38
rs951533342 Chr.2: 168930773-168930773 on Build GRCh38
rs919455454 Chr.2: 168930845-168930845 on Build GRCh38
rs11568359 Chr.2: 168930818-168930818 on Build GRCh38
rs377222158 Chr.2: 168930766-168930766 on Build GRCh38
rs377593604 Chr.2: 168930869-168930869 on Build GRCh38
rs750944055 Chr.2: 168930830-168930830 on Build GRCh38
rs929482399 Chr.2: 168930848-168930848 on Build GRCh38
rs764581483 Chr.2: 168930694-168930694 on Build GRCh38
rs72549396 Chr.2: 168930808-168930808 on Build GRCh38
rs373201488 Chr.2: 168930843-168930843 on Build GRCh38
rs1031540522 Chr.2: 168930674-168930674 on Build GRCh38
rs1030004521 Chr.2: 168930798-168930798 on Build GRCh38
rs891695981 Chr.2: 168930735-168930735 on Build GRCh38
rs762737233 Chr.2: 168930771-168930771 on Build GRCh38
rs377691925 Chr.2: 168930770-168930770 on Build GRCh38
rs747920364 Chr.2: 168930759-168930759 on Build GRCh38
rs768455510 Chr.2: 168930650-168930650 on Build GRCh38
rs750991541 Chr.2: 168930868-168930868 on Build GRCh38
rs756220860 Chr.2: 168930693-168930693 on Build GRCh38
rs757887126 Chr.2: 168930701-168930701 on Build GRCh38
rs754048936 Chr.2: 168930726-168930726 on Build GRCh38

View Full Product Details