Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6916943-6917204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO2
Entrez Gene ID
2026
Gene Name
enolase 2
Gene Aliases
HEL-S-279, NSE
UniGene
Hs.511915
Gene Chromosome Location
Chr.12: 6914450-6923696 on Build GRCh38
Transcripts
NM_001975.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5906937 c.262G>A p.E88K Chr.12: 6917059-6917059 on Build GRCh38
4400986 c.263A>T p.E88V Chr.12: 6917060-6917060 on Build GRCh38
7289960 c.241-1G>T p.? Chr.12: 6917037-6917037 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs782575715 Chr.12: 6917147-6917147 on Build GRCh38
rs376565830 Chr.12: 6917086-6917086 on Build GRCh38
rs1052229003 Chr.12: 6917002-6917002 on Build GRCh38
rs782283343 Chr.12: 6917115-6917115 on Build GRCh38
rs372023195 Chr.12: 6917121-6917121 on Build GRCh38
rs781851933 Chr.12: 6917050-6917050 on Build GRCh38
rs782686386 Chr.12: 6917114-6917114 on Build GRCh38
rs782755928 Chr.12: 6917154-6917154 on Build GRCh38
rs782234692 Chr.12: 6917009-6917009 on Build GRCh38
rs544881867 Chr.12: 6916982-6916982 on Build GRCh38
rs925208601 Chr.12: 6916975-6916975 on Build GRCh38
rs781994800 Chr.12: 6917134-6917134 on Build GRCh38
rs782080778 Chr.12: 6917041-6917041 on Build GRCh38
rs2071417 Chr.12: 6917143-6917143 on Build GRCh38
rs781788785 Chr.12: 6917180-6917180 on Build GRCh38
rs200824186 Chr.12: 6917135-6917135 on Build GRCh38
rs373379279 Chr.12: 6917128-6917128 on Build GRCh38
rs782790445 Chr.12: 6917058-6917058 on Build GRCh38
rs781958410 Chr.12: 6917145-6917145 on Build GRCh38
rs192215364 Chr.12: 6917139-6917139 on Build GRCh38
rs782337922 Chr.12: 6917184-6917184 on Build GRCh38
rs782615624 Chr.12: 6917091-6917091 on Build GRCh38
rs782249919 Chr.12: 6917019-6917019 on Build GRCh38
rs140873913 Chr.12: 6917136-6917136 on Build GRCh38
rs781876639 Chr.12: 6917072-6917072 on Build GRCh38
rs782405237 Chr.12: 6917020-6917020 on Build GRCh38
rs782486777 Chr.12: 6917073-6917073 on Build GRCh38
rs564575090 Chr.12: 6917120-6917120 on Build GRCh38
rs782773921 Chr.12: 6917043-6917043 on Build GRCh38
rs782012005 Chr.12: 6917138-6917138 on Build GRCh38
rs782303286 Chr.12: 6917031-6917031 on Build GRCh38
rs782060480 Chr.12: 6917028-6917028 on Build GRCh38
rs782541357 Chr.12: 6917160-6917160 on Build GRCh38
rs782716149 Chr.12: 6917169-6917169 on Build GRCh38
rs368918497 Chr.12: 6917103-6917103 on Build GRCh38
rs782381949 Chr.12: 6917018-6917019 on Build GRCh38
rs12320576 Chr.12: 6916980-6916980 on Build GRCh38
rs782162349 Chr.12: 6917052-6917052 on Build GRCh38
rs184095556 Chr.12: 6917155-6917155 on Build GRCh38
rs782651799 Chr.12: 6917016-6917016 on Build GRCh38
rs782270453 Chr.12: 6917003-6917003 on Build GRCh38
rs782336478 Chr.12: 6917132-6917132 on Build GRCh38
rs782506378 Chr.12: 6917107-6917107 on Build GRCh38
rs373941121 Chr.12: 6917024-6917024 on Build GRCh38
rs369056037 Chr.12: 6917014-6917014 on Build GRCh38

View Full Product Details