Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 151438153-151438422 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGSF10
Entrez Gene ID
285313
Gene Name
immunoglobulin superfamily member 10
Gene Aliases
CMF608
UniGene
Hs.58561
Gene Chromosome Location
Chr.3: 151435051-151619926 on Build GRCh38
Transcripts
NM_001178145.1 NM_001178146.1 NM_178822.4 XM_011512708.2 XM_011512709.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3915156 c.6390G>A p.M2130I Chr.3: 151438171-151438171 on Build GRCh38
6578034 c.6212C>A p.S2071Y Chr.3: 151438349-151438349 on Build GRCh38
4896478 c.6157G>A p.G2053R Chr.3: 151438404-151438404 on Build GRCh38
5891753 c.6259G>A p.D2087N Chr.3: 151438302-151438302 on Build GRCh38
6572768 c.6140G>C p.R2047T Chr.3: 151438421-151438421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746593123 Chr.3: 151438362-151438362 on Build GRCh38
rs377142937 Chr.3: 151438226-151438226 on Build GRCh38
rs778522469 Chr.3: 151438224-151438224 on Build GRCh38
rs754946545 Chr.3: 151438230-151438230 on Build GRCh38
rs527825904 Chr.3: 151438324-151438324 on Build GRCh38
rs771352318 Chr.3: 151438344-151438344 on Build GRCh38
rs781113596 Chr.3: 151438318-151438318 on Build GRCh38
rs1051552779 Chr.3: 151438382-151438382 on Build GRCh38
rs771834714 Chr.3: 151438271-151438271 on Build GRCh38
rs764102480 Chr.3: 151438218-151438218 on Build GRCh38
rs886431262 Chr.3: 151438350-151438350 on Build GRCh38
rs767970971 Chr.3: 151438235-151438235 on Build GRCh38
rs138856486 Chr.3: 151438257-151438257 on Build GRCh38
rs751741616 Chr.3: 151438310-151438310 on Build GRCh38
rs374514917 Chr.3: 151438384-151438384 on Build GRCh38
rs752247874 Chr.3: 151438210-151438210 on Build GRCh38
rs768826828 Chr.3: 151438215-151438217 on Build GRCh38
rs374148063 Chr.3: 151438338-151438338 on Build GRCh38
rs760750544 Chr.3: 151438215-151438215 on Build GRCh38
rs747106942 Chr.3: 151438228-151438228 on Build GRCh38
rs140636759 Chr.3: 151438285-151438285 on Build GRCh38
rs78115932 Chr.3: 151438225-151438225 on Build GRCh38
rs145634427 Chr.3: 151438316-151438316 on Build GRCh38
rs758133735 Chr.3: 151438305-151438305 on Build GRCh38
rs75202100 Chr.3: 151438308-151438308 on Build GRCh38
rs768022610 Chr.3: 151438387-151438387 on Build GRCh38
rs753647662 Chr.3: 151438221-151438221 on Build GRCh38
rs775309595 Chr.3: 151438353-151438353 on Build GRCh38
rs982493163 Chr.3: 151438258-151438258 on Build GRCh38
rs912530027 Chr.3: 151438386-151438386 on Build GRCh38
rs200998219 Chr.3: 151438222-151438222 on Build GRCh38
rs761633551 Chr.3: 151438298-151438298 on Build GRCh38
rs370277399 Chr.3: 151438247-151438247 on Build GRCh38
rs776651746 Chr.3: 151438292-151438292 on Build GRCh38
rs975322538 Chr.3: 151438246-151438246 on Build GRCh38
rs908711998 Chr.3: 151438277-151438277 on Build GRCh38
rs767178288 Chr.3: 151438202-151438202 on Build GRCh38
rs768260632 Chr.3: 151438371-151438371 on Build GRCh38
rs368664622 Chr.3: 151438372-151438372 on Build GRCh38
rs760233090 Chr.3: 151438279-151438279 on Build GRCh38
rs749681801 Chr.3: 151438342-151438342 on Build GRCh38
rs748559170 Chr.3: 151438236-151438236 on Build GRCh38
rs750208612 Chr.3: 151438303-151438303 on Build GRCh38
rs201909176 Chr.3: 151438302-151438302 on Build GRCh38
rs775221370 Chr.3: 151438273-151438273 on Build GRCh38
rs761874663 Chr.3: 151438381-151438381 on Build GRCh38
rs114334639 Chr.3: 151438193-151438193 on Build GRCh38
rs776591826 Chr.3: 151438258-151438259 on Build GRCh38
rs150254783 Chr.3: 151438231-151438231 on Build GRCh38
rs922497375 Chr.3: 151438261-151438261 on Build GRCh38
rs773684022 Chr.3: 151438253-151438253 on Build GRCh38

View Full Product Details