Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1338397-1338662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DVL1
Entrez Gene ID
1855
Gene Name
dishevelled segment polarity protein 1
Gene Aliases
DRS2, DVL, DVL1L1, DVL1P1
UniGene
Hs.731450
Gene Chromosome Location
Chr.1: 1335278-1349142 on Build GRCh38
Transcripts
NM_004421.2 XM_005244731.3 XM_005244732.3 XM_005244733.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6650446 c.1297G>A p.D433N Chr.1: 1338564-1338564 on Build GRCh38
4679755 c.1348G>A p.V450M Chr.1: 1338428-1338428 on Build GRCh38
1579345 c.1134G>T p.E378D Chr.1: 1338652-1338652 on Build GRCh38
7171587 c.1240G>A p.D414N Chr.1: 1338621-1338621 on Build GRCh38
4021440 c.1226G>A p.R409H Chr.1: 1338560-1338560 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144133102 Chr.1: 1338640-1338640 on Build GRCh38
rs1025311770 Chr.1: 1338500-1338500 on Build GRCh38
rs756786864 Chr.1: 1338617-1338617 on Build GRCh38
rs141420415 Chr.1: 1338583-1338583 on Build GRCh38
rs778631183 Chr.1: 1338496-1338496 on Build GRCh38
rs148871455 Chr.1: 1338580-1338580 on Build GRCh38
rs755815862 Chr.1: 1338489-1338489 on Build GRCh38
rs774744891 Chr.1: 1338637-1338637 on Build GRCh38
rs768207125 Chr.1: 1338429-1338429 on Build GRCh38
rs370619119 Chr.1: 1338503-1338503 on Build GRCh38
rs765147049 Chr.1: 1338600-1338600 on Build GRCh38
rs756176832 Chr.1: 1338533-1338533 on Build GRCh38
rs372788630 Chr.1: 1338603-1338603 on Build GRCh38
rs937427372 Chr.1: 1338469-1338469 on Build GRCh38
rs374474114 Chr.1: 1338607-1338607 on Build GRCh38
rs746424149 Chr.1: 1338621-1338621 on Build GRCh38
rs747583654 Chr.1: 1338506-1338506 on Build GRCh38
rs571199278 Chr.1: 1338591-1338591 on Build GRCh38
rs764307176 Chr.1: 1338522-1338522 on Build GRCh38
rs199890964 Chr.1: 1338468-1338468 on Build GRCh38
rs776830656 Chr.1: 1338461-1338461 on Build GRCh38
rs932467296 Chr.1: 1338455-1338455 on Build GRCh38
rs770046620 Chr.1: 1338622-1338622 on Build GRCh38
rs777325356 Chr.1: 1338565-1338565 on Build GRCh38
rs750431968 Chr.1: 1338478-1338478 on Build GRCh38
rs867144796 Chr.1: 1338432-1338432 on Build GRCh38
rs777410024 Chr.1: 1338609-1338609 on Build GRCh38
rs1035970545 Chr.1: 1338501-1338501 on Build GRCh38
rs375802354 Chr.1: 1338638-1338638 on Build GRCh38
rs1026836522 Chr.1: 1338505-1338505 on Build GRCh38
rs765342412 Chr.1: 1338512-1338512 on Build GRCh38
rs1053500069 Chr.1: 1338477-1338477 on Build GRCh38
rs759220527 Chr.1: 1338599-1338599 on Build GRCh38
rs747707366 Chr.1: 1338443-1338443 on Build GRCh38
rs767509229 Chr.1: 1338473-1338473 on Build GRCh38
rs183970155 Chr.1: 1338490-1338490 on Build GRCh38
rs773789094 Chr.1: 1338433-1338433 on Build GRCh38
rs759737026 Chr.1: 1338467-1338467 on Build GRCh38
rs776926808 Chr.1: 1338508-1338508 on Build GRCh38
rs773679400 Chr.1: 1338504-1338504 on Build GRCh38
rs753509462 Chr.1: 1338538-1338538 on Build GRCh38
rs781525302 Chr.1: 1338618-1338618 on Build GRCh38
rs780523329 Chr.1: 1338630-1338630 on Build GRCh38
rs748985130 Chr.1: 1338428-1338428 on Build GRCh38
rs186806059 Chr.1: 1338494-1338494 on Build GRCh38
rs766432002 Chr.1: 1338537-1338537 on Build GRCh38
rs775799425 Chr.1: 1338513-1338513 on Build GRCh38
rs937376607 Chr.1: 1338628-1338628 on Build GRCh38
rs376201539 Chr.1: 1338451-1338451 on Build GRCh38
rs752434991 Chr.1: 1338558-1338558 on Build GRCh38
rs750379091 Chr.1: 1338529-1338529 on Build GRCh38
rs897096552 Chr.1: 1338502-1338502 on Build GRCh38
rs371482747 Chr.1: 1338631-1338631 on Build GRCh38
rs773730913 Chr.1: 1338452-1338452 on Build GRCh38
rs759953229 Chr.1: 1338511-1338511 on Build GRCh38
rs753835995 Chr.1: 1338598-1338598 on Build GRCh38
rs893243731 Chr.1: 1338489-1338489 on Build GRCh38
rs144876679 Chr.1: 1338541-1338541 on Build GRCh38
rs762915558 Chr.1: 1338515-1338515 on Build GRCh38
rs755938775 Chr.1: 1338480-1338480 on Build GRCh38
rs746537493 Chr.1: 1338567-1338567 on Build GRCh38
rs761987800 Chr.1: 1338587-1338587 on Build GRCh38
rs1051356744 Chr.1: 1338606-1338606 on Build GRCh38
rs375510995 Chr.1: 1338420-1338420 on Build GRCh38
rs1057518627 Chr.1: 1338529-1338530 on Build GRCh38
rs534728716 Chr.1: 1338571-1338571 on Build GRCh38
rs138293356 Chr.1: 1338639-1338639 on Build GRCh38
rs757900986 Chr.1: 1338564-1338564 on Build GRCh38
rs768931734 Chr.1: 1338632-1338632 on Build GRCh38
rs952424082 Chr.1: 1338509-1338509 on Build GRCh38
rs770238250 Chr.1: 1338576-1338576 on Build GRCh38
rs369983320 Chr.1: 1338460-1338460 on Build GRCh38
rs998894703 Chr.1: 1338476-1338476 on Build GRCh38
rs201616962 Chr.1: 1338579-1338579 on Build GRCh38
rs778408006 Chr.1: 1338557-1338557 on Build GRCh38
rs111891916 Chr.1: 1338499-1338499 on Build GRCh38
rs867732546 Chr.1: 1338560-1338560 on Build GRCh38
rs375888432 Chr.1: 1338568-1338568 on Build GRCh38
rs751033868 Chr.1: 1338610-1338610 on Build GRCh38
rs780058154 Chr.1: 1338485-1338485 on Build GRCh38

View Full Product Details