Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 14747005-14747266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FREM1
Entrez Gene ID
158326
Gene Name
FRAS1 related extracellular matrix 1
Gene Aliases
BNAR, C9orf143, C9orf145, C9orf154, MOTA, TILRR, TRIGNO2
UniGene
Hs.50850
Gene Chromosome Location
Chr.9: 14734666-14911653 on Build GRCh38
Transcripts
NM_001177704.1 NM_144966.5 XM_005251382.3 XM_005251384.4 XM_006716729.3 XM_011517758.2 XM_017014316.1 XM_017014317.1 XM_017014318.1 XM_017014319.1 XM_017014320.1 XM_017014321.1 XM_017014322.1 XM_017014323.1 XM_017014324.1 XM_017014325.1 XM_017014326.1 XM_017014327.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5410855 c.6015G>A p.Q2005Q Chr.9: 14747046-14747046 on Build GRCh38
5410857 c.6010G>A p.D2004N Chr.9: 14747051-14747051 on Build GRCh38
7092808 c.1641G>T p.K547N Chr.9: 14747028-14747028 on Build GRCh38
3926533 c.1663G>A p.G555R Chr.9: 14747006-14747006 on Build GRCh38
5968094 c.6054G>T p.K2018N Chr.9: 14747007-14747007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762234673 Chr.9: 14747100-14747100 on Build GRCh38
rs932684585 Chr.9: 14747104-14747104 on Build GRCh38
rs763196871 Chr.9: 14747234-14747237 on Build GRCh38
rs778888575 Chr.9: 14747062-14747062 on Build GRCh38
rs757721445 Chr.9: 14747188-14747188 on Build GRCh38
rs1005547572 Chr.9: 14747132-14747132 on Build GRCh38
rs746604828 Chr.9: 14747078-14747078 on Build GRCh38
rs756420258 Chr.9: 14747031-14747031 on Build GRCh38
rs200987019 Chr.9: 14747040-14747040 on Build GRCh38
rs774868755 Chr.9: 14747096-14747096 on Build GRCh38
rs776329304 Chr.9: 14747090-14747090 on Build GRCh38
rs754058350 Chr.9: 14747233-14747233 on Build GRCh38
rs544661735 Chr.9: 14747219-14747219 on Build GRCh38
rs1049766490 Chr.9: 14747106-14747106 on Build GRCh38
rs990899832 Chr.9: 14747076-14747076 on Build GRCh38
rs777270214 Chr.9: 14747075-14747075 on Build GRCh38
rs578046556 Chr.9: 14747086-14747086 on Build GRCh38
rs1053579583 Chr.9: 14747170-14747170 on Build GRCh38
rs2270530 Chr.9: 14747159-14747159 on Build GRCh38
rs200035180 Chr.9: 14747235-14747235 on Build GRCh38
rs2270529 Chr.9: 14747135-14747135 on Build GRCh38
rs889711285 Chr.9: 14747174-14747174 on Build GRCh38
rs748058742 Chr.9: 14747064-14747064 on Build GRCh38
rs572603801 Chr.9: 14747209-14747209 on Build GRCh38
rs949196536 Chr.9: 14747087-14747087 on Build GRCh38
rs1016487209 Chr.9: 14747067-14747067 on Build GRCh38
rs558194682 Chr.9: 14747073-14747073 on Build GRCh38
rs917583009 Chr.9: 14747082-14747082 on Build GRCh38
rs542797406 Chr.9: 14747165-14747165 on Build GRCh38
rs749631410 Chr.9: 14747091-14747091 on Build GRCh38
rs888482142 Chr.9: 14747128-14747128 on Build GRCh38
rs191713464 Chr.9: 14747054-14747054 on Build GRCh38
rs73644845 Chr.9: 14747180-14747180 on Build GRCh38
rs1039486407 Chr.9: 14747203-14747203 on Build GRCh38
rs145607985 Chr.9: 14747136-14747136 on Build GRCh38
rs564553123 Chr.9: 14747234-14747234 on Build GRCh38
rs934520223 Chr.9: 14747033-14747033 on Build GRCh38
rs1004132383 Chr.9: 14747138-14747138 on Build GRCh38
rs566175340 Chr.9: 14747052-14747052 on Build GRCh38
rs375371745 Chr.9: 14747216-14747216 on Build GRCh38
rs766678522 Chr.9: 14747230-14747230 on Build GRCh38

View Full Product Details