Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 101723547-101723776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHPT1
Entrez Gene ID
56994
Gene Name
choline phosphotransferase 1
Gene Aliases
CPT, CPT1
UniGene
Hs.293077
Gene Chromosome Location
Chr.12: 101697639-101752038 on Build GRCh38
Transcripts
NM_020244.2 XM_011538574.1 XM_011538575.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
430171 c.973C>T p.Q325* Chr.12: 101723755-101723755 on Build GRCh38
3667074 c.968A>G p.Y323C Chr.12: 101723750-101723750 on Build GRCh38
3416387 c.956A>G p.K319R Chr.12: 101723738-101723738 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764915335 Chr.12: 101723702-101723705 on Build GRCh38
rs757929652 Chr.12: 101723742-101723742 on Build GRCh38
rs1012925370 Chr.12: 101723588-101723588 on Build GRCh38
rs756911163 Chr.12: 101723673-101723674 on Build GRCh38
rs776680337 Chr.12: 101723697-101723697 on Build GRCh38
rs761803831 Chr.12: 101723701-101723701 on Build GRCh38
rs569832616 Chr.12: 101723663-101723663 on Build GRCh38
rs1028571092 Chr.12: 101723670-101723670 on Build GRCh38
rs577799309 Chr.12: 101723585-101723585 on Build GRCh38
rs1040473612 Chr.12: 101723590-101723590 on Build GRCh38
rs555795326 Chr.12: 101723636-101723636 on Build GRCh38
rs971654599 Chr.12: 101723619-101723619 on Build GRCh38
rs368918351 Chr.12: 101723584-101723584 on Build GRCh38
rs776461364 Chr.12: 101723625-101723629 on Build GRCh38
rs537286227 Chr.12: 101723607-101723607 on Build GRCh38
rs541777969 Chr.12: 101723716-101723716 on Build GRCh38
rs765290675 Chr.12: 101723709-101723709 on Build GRCh38
rs535029486 Chr.12: 101723575-101723575 on Build GRCh38
rs900548306 Chr.12: 101723662-101723662 on Build GRCh38
rs1023200736 Chr.12: 101723592-101723592 on Build GRCh38
rs750040817 Chr.12: 101723711-101723711 on Build GRCh38
rs764029502 Chr.12: 101723694-101723694 on Build GRCh38
rs751634924 Chr.12: 101723731-101723731 on Build GRCh38

View Full Product Details