Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 48878219-48878468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EID1
Entrez Gene ID
23741
Gene Name
EP300 interacting inhibitor of differentiation 1
Gene Aliases
C15orf3, CRI1, EID-1, IRO45620, PNAS-22, PTD014, RBP21
UniGene
Hs.255973
Gene Chromosome Location
Chr.15: 48878093-48880183 on Build GRCh38
Transcripts
NM_014335.2 NM_203349.3 XM_005254375.3

Gene Symbol
SHC4
Entrez Gene ID
399694
Gene Name
SHC adaptor protein 4
Gene Aliases
RaLP, SHCD
UniGene
Hs.642615
Gene Chromosome Location
Chr.15: 48823737-48963444 on Build GRCh38
Transcripts
NM_014335.2 NM_203349.3 XM_005254375.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5180072 c.143G>A p.G48E Chr.15: 48878319-48878319 on Build GRCh38
6194967 c.145G>C p.A49P Chr.15: 48878321-48878321 on Build GRCh38
1587908 c.157G>A p.E53K Chr.15: 48878333-48878333 on Build GRCh38
5095204 c.239A>T p.N80I Chr.15: 48878415-48878415 on Build GRCh38
470741 c.123C>A p.S41S Chr.15: 48878299-48878299 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764215628 Chr.15: 48878444-48878444 on Build GRCh38
rs761502658 Chr.15: 48878408-48878408 on Build GRCh38
rs746356498 Chr.15: 48878321-48878321 on Build GRCh38
rs761401009 Chr.15: 48878346-48878346 on Build GRCh38
rs766200338 Chr.15: 48878349-48878350 on Build GRCh38
rs772982315 Chr.15: 48878414-48878414 on Build GRCh38
rs775135586 Chr.15: 48878326-48878326 on Build GRCh38
rs774881936 Chr.15: 48878319-48878319 on Build GRCh38
rs1028481351 Chr.15: 48878436-48878436 on Build GRCh38
rs780651046 Chr.15: 48878397-48878397 on Build GRCh38
rs370992223 Chr.15: 48878248-48878248 on Build GRCh38
rs562466685 Chr.15: 48878372-48878372 on Build GRCh38
rs532770133 Chr.15: 48878394-48878394 on Build GRCh38
rs370518786 Chr.15: 48878383-48878383 on Build GRCh38
rs757605087 Chr.15: 48878379-48878379 on Build GRCh38
rs932959866 Chr.15: 48878322-48878322 on Build GRCh38
rs11559087 Chr.15: 48878261-48878261 on Build GRCh38
rs750319038 Chr.15: 48878275-48878275 on Build GRCh38
rs752702144 Chr.15: 48878361-48878361 on Build GRCh38
rs753562915 Chr.15: 48878253-48878253 on Build GRCh38
rs758350829 Chr.15: 48878294-48878294 on Build GRCh38
rs867896701 Chr.15: 48878261-48878262 on Build GRCh38
rs540468266 Chr.15: 48878240-48878240 on Build GRCh38
rs977129684 Chr.15: 48878302-48878302 on Build GRCh38
rs776024788 Chr.15: 48878405-48878405 on Build GRCh38
rs757018335 Chr.15: 48878266-48878266 on Build GRCh38
rs750264190 Chr.15: 48878357-48878357 on Build GRCh38
rs768064540 Chr.15: 48878332-48878332 on Build GRCh38
rs566866767 Chr.15: 48878442-48878442 on Build GRCh38
rs769474048 Chr.15: 48878409-48878409 on Build GRCh38
rs762766855 Chr.15: 48878350-48878352 on Build GRCh38
rs748264256 Chr.15: 48878310-48878310 on Build GRCh38
rs747474162 Chr.15: 48878402-48878402 on Build GRCh38
rs766365825 Chr.15: 48878360-48878360 on Build GRCh38
rs771290460 Chr.15: 48878318-48878318 on Build GRCh38
rs377583239 Chr.15: 48878335-48878335 on Build GRCh38
rs374971624 Chr.15: 48878393-48878393 on Build GRCh38
rs754045355 Chr.15: 48878374-48878374 on Build GRCh38
rs768172432 Chr.15: 48878404-48878404 on Build GRCh38
rs760215753 Chr.15: 48878241-48878241 on Build GRCh38
rs759416600 Chr.15: 48878438-48878438 on Build GRCh38
rs751490550 Chr.15: 48878386-48878387 on Build GRCh38
rs753906069 Chr.15: 48878450-48878450 on Build GRCh38
rs543988670 Chr.15: 48878359-48878359 on Build GRCh38
rs551657799 Chr.15: 48878413-48878413 on Build GRCh38
rs561606415 Chr.15: 48878308-48878308 on Build GRCh38
rs751478270 Chr.15: 48878421-48878421 on Build GRCh38
rs375455351 Chr.15: 48878317-48878317 on Build GRCh38
rs762807833 Chr.15: 48878313-48878313 on Build GRCh38
rs756155052 Chr.15: 48878365-48878365 on Build GRCh38
rs777997601 Chr.15: 48878315-48878315 on Build GRCh38
rs528956826 Chr.15: 48878329-48878329 on Build GRCh38

View Full Product Details