Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 58328026-58328299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID4A
Entrez Gene ID
5926
Gene Name
AT-rich interaction domain 4A
Gene Aliases
RBBP-1, RBBP1, RBP-1, RBP1
UniGene
Hs.161000
Gene Chromosome Location
Chr.14: 58298385-58373733 on Build GRCh38
Transcripts
NM_002892.3 NM_023000.2 NM_023001.2 XM_005267964.2 XM_005267965.2 XM_017021560.1 XM_017021561.1 XM_017021562.1 XM_017021563.1 XM_017021564.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5027865 c.591T>G p.S197S Chr.14: 58328245-58328245 on Build GRCh38
6604670 c.641G>A p.R214Q Chr.14: 58328295-58328295 on Build GRCh38
5596291 c.644C>T p.S215L Chr.14: 58328298-58328298 on Build GRCh38
5757826 c.585A>G p.V195V Chr.14: 58328239-58328239 on Build GRCh38
192773 c.640C>T p.R214* Chr.14: 58328294-58328294 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs537071810 Chr.14: 58328136-58328136 on Build GRCh38
rs950640177 Chr.14: 58328125-58328125 on Build GRCh38
rs745415308 Chr.14: 58328265-58328265 on Build GRCh38
rs1025589606 Chr.14: 58328078-58328078 on Build GRCh38
rs369499130 Chr.14: 58328261-58328261 on Build GRCh38
rs75887271 Chr.14: 58328212-58328212 on Build GRCh38
rs375234834 Chr.14: 58328213-58328213 on Build GRCh38
rs1026221068 Chr.14: 58328094-58328094 on Build GRCh38
rs994388537 Chr.14: 58328072-58328072 on Build GRCh38
rs576095119 Chr.14: 58328132-58328132 on Build GRCh38
rs757236554 Chr.14: 58328260-58328260 on Build GRCh38
rs777615225 Chr.14: 58328254-58328254 on Build GRCh38
rs755910554 Chr.14: 58328250-58328250 on Build GRCh38
rs75756700 Chr.14: 58328082-58328082 on Build GRCh38
rs781741745 Chr.14: 58328051-58328051 on Build GRCh38
rs767806287 Chr.14: 58328221-58328221 on Build GRCh38
rs911183441 Chr.14: 58328197-58328197 on Build GRCh38
rs986781868 Chr.14: 58328148-58328148 on Build GRCh38
rs752808395 Chr.14: 58328224-58328224 on Build GRCh38
rs536546893 Chr.14: 58328059-58328059 on Build GRCh38
rs749071631 Chr.14: 58328257-58328257 on Build GRCh38
rs754570853 Chr.14: 58328256-58328258 on Build GRCh38
rs771424991 Chr.14: 58328267-58328267 on Build GRCh38
rs746829211 Chr.14: 58328228-58328229 on Build GRCh38
rs962612407 Chr.14: 58328064-58328064 on Build GRCh38
rs917058566 Chr.14: 58328147-58328147 on Build GRCh38
rs1017086729 Chr.14: 58328058-58328058 on Build GRCh38
rs992659126 Chr.14: 58328142-58328142 on Build GRCh38

View Full Product Details