Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 69586117-69586390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT2
Entrez Gene ID
10576
Gene Name
chaperonin containing TCP1 subunit 2
Gene Aliases
99D8.1, CCT-beta, CCTB, HEL-S-100n, PRO1633, TCP-1-beta
UniGene
Hs.189772
Gene Chromosome Location
Chr.12: 69585428-69601577 on Build GRCh38
Transcripts
NM_001198842.1 NM_006431.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5375861 c.9C>T p.S3S Chr.12: 69586275-69586275 on Build GRCh38
6462888 c.4-4T>C p.? Chr.12: 69586266-69586266 on Build GRCh38
3464587 c.8C>T p.S3F Chr.12: 69586274-69586274 on Build GRCh38
6392805 c.71C>A p.A24D Chr.12: 69586337-69586337 on Build GRCh38
6623281 c.45A>G p.G15G Chr.12: 69586311-69586311 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs373807219 Chr.12: 69586271-69586271 on Build GRCh38
rs751339085 Chr.12: 69586290-69586290 on Build GRCh38
rs1040359309 Chr.12: 69586317-69586317 on Build GRCh38
rs367696863 Chr.12: 69586288-69586288 on Build GRCh38
rs749771956 Chr.12: 69586357-69586357 on Build GRCh38
rs755206069 Chr.12: 69586296-69586296 on Build GRCh38
rs762762710 Chr.12: 69586249-69586249 on Build GRCh38
rs484319 Chr.12: 69586361-69586361 on Build GRCh38
rs752722597 Chr.12: 69586304-69586304 on Build GRCh38
rs111309939 Chr.12: 69586302-69586302 on Build GRCh38
rs768854556 Chr.12: 69586223-69586224 on Build GRCh38
rs150207506 Chr.12: 69586236-69586236 on Build GRCh38
rs914410285 Chr.12: 69586217-69586217 on Build GRCh38
rs561022013 Chr.12: 69586153-69586155 on Build GRCh38
rs200874735 Chr.12: 69586231-69586231 on Build GRCh38
rs907524811 Chr.12: 69586153-69586153 on Build GRCh38
rs748905110 Chr.12: 69586209-69586209 on Build GRCh38
rs778447483 Chr.12: 69586353-69586353 on Build GRCh38
rs11541104 Chr.12: 69586309-69586309 on Build GRCh38
rs485288 Chr.12: 69586248-69586248 on Build GRCh38
rs765166456 Chr.12: 69586247-69586247 on Build GRCh38
rs371275276 Chr.12: 69586215-69586215 on Build GRCh38
rs988189241 Chr.12: 69586311-69586311 on Build GRCh38
rs766123831 Chr.12: 69586280-69586280 on Build GRCh38
rs981185830 Chr.12: 69586205-69586205 on Build GRCh38
rs993856677 Chr.12: 69586189-69586189 on Build GRCh38
rs756203347 Chr.12: 69586306-69586306 on Build GRCh38
rs776059523 Chr.12: 69586242-69586242 on Build GRCh38
rs964312701 Chr.12: 69586255-69586255 on Build GRCh38
rs568746707 Chr.12: 69586212-69586212 on Build GRCh38
rs775238651 Chr.12: 69586226-69586226 on Build GRCh38
rs776742327 Chr.12: 69586318-69586320 on Build GRCh38
rs761378029 Chr.12: 69586246-69586246 on Build GRCh38
rs570539360 Chr.12: 69586152-69586152 on Build GRCh38

View Full Product Details