Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 62320357-62320624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf14
Entrez Gene ID
57415
Gene Name
chromosome 3 open reading frame 14
Gene Aliases
HT021
UniGene
Hs.47166
Gene Chromosome Location
Chr.3: 62318973-62336213 on Build GRCh38
Transcripts
NM_001291941.1 NM_001291942.1 NM_001291943.1 NM_020685.4 XM_017006931.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6615982 c.53+5_53+6insT p.? Chr.3: 62320529-62320530 on Build GRCh38
5465947 c.28C>T p.R10C Chr.3: 62320499-62320499 on Build GRCh38
6615983 c.47A>C p.E16A Chr.3: 62320518-62320518 on Build GRCh38
1176636 c.35C>A p.S12Y Chr.3: 62320506-62320506 on Build GRCh38
7171243 c.16G>A p.A6T Chr.3: 62320487-62320487 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1041438256 Chr.3: 62320385-62320385 on Build GRCh38
rs142011355 Chr.3: 62320499-62320499 on Build GRCh38
rs945502406 Chr.3: 62320557-62320557 on Build GRCh38
rs924477754 Chr.3: 62320575-62320575 on Build GRCh38
rs753441367 Chr.3: 62320441-62320441 on Build GRCh38
rs375637308 Chr.3: 62320484-62320484 on Build GRCh38
rs910800439 Chr.3: 62320466-62320466 on Build GRCh38
rs112502637 Chr.3: 62320463-62320463 on Build GRCh38
rs748738206 Chr.3: 62320525-62320525 on Build GRCh38
rs777263427 Chr.3: 62320432-62320433 on Build GRCh38
rs770105664 Chr.3: 62320538-62320538 on Build GRCh38
rs760171871 Chr.3: 62320422-62320422 on Build GRCh38
rs778287844 Chr.3: 62320496-62320496 on Build GRCh38
rs775640530 Chr.3: 62320549-62320549 on Build GRCh38
rs745989768 Chr.3: 62320532-62320532 on Build GRCh38
rs754603542 Chr.3: 62320450-62320450 on Build GRCh38
rs765907236 Chr.3: 62320440-62320440 on Build GRCh38
rs139980298 Chr.3: 62320500-62320500 on Build GRCh38
rs764909915 Chr.3: 62320460-62320460 on Build GRCh38
rs902869459 Chr.3: 62320391-62320391 on Build GRCh38
rs372692739 Chr.3: 62320559-62320559 on Build GRCh38
rs771776438 Chr.3: 62320566-62320566 on Build GRCh38
rs1024244540 Chr.3: 62320520-62320520 on Build GRCh38
rs1012496690 Chr.3: 62320512-62320512 on Build GRCh38
rs773408861 Chr.3: 62320507-62320507 on Build GRCh38
rs915351497 Chr.3: 62320522-62320522 on Build GRCh38
rs770022165 Chr.3: 62320530-62320530 on Build GRCh38
rs527736726 Chr.3: 62320529-62320529 on Build GRCh38
rs999822966 Chr.3: 62320432-62320432 on Build GRCh38
rs962023631 Chr.3: 62320544-62320544 on Build GRCh38
rs753105297 Chr.3: 62320483-62320483 on Build GRCh38
rs989428492 Chr.3: 62320487-62320487 on Build GRCh38
rs773132977 Chr.3: 62320568-62320568 on Build GRCh38
rs760629271 Chr.3: 62320574-62320574 on Build GRCh38

View Full Product Details