Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 20764896-20765131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOCAD
Entrez Gene ID
54914
Gene Name
focadhesin
Gene Aliases
KIAA1797
UniGene
Hs.136247
Gene Chromosome Location
Chr.9: 20658309-20995955 on Build GRCh38
Transcripts
NM_017794.4 XM_005251494.3 XM_011517945.2 XM_017014852.1 XM_017014853.1 XM_017014854.1 XM_017014855.1 XM_017014856.1 XM_017014857.1 XM_017014858.1 XM_017014859.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
753797 c.534G>T p.L178L Chr.9: 20764908-20764908 on Build GRCh38
4854271 c.568G>T p.E190* Chr.9: 20764942-20764942 on Build GRCh38
6115620 c.532C>G p.L178V Chr.9: 20764906-20764906 on Build GRCh38
5545010 c.689C>T p.P230L Chr.9: 20765063-20765063 on Build GRCh38
4499422 c.541C>T p.L181L Chr.9: 20764915-20764915 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs7871811 Chr.9: 20765098-20765098 on Build GRCh38
rs536812144 Chr.9: 20764961-20764961 on Build GRCh38
rs897351146 Chr.9: 20765093-20765093 on Build GRCh38
rs772604881 Chr.9: 20764956-20764956 on Build GRCh38
rs912644058 Chr.9: 20764987-20764987 on Build GRCh38
rs769073717 Chr.9: 20764946-20764946 on Build GRCh38
rs779646531 Chr.9: 20765008-20765008 on Build GRCh38
rs747448535 Chr.9: 20765026-20765026 on Build GRCh38
rs780960707 Chr.9: 20764938-20764938 on Build GRCh38
rs750105844 Chr.9: 20765004-20765004 on Build GRCh38
rs762899573 Chr.9: 20765051-20765051 on Build GRCh38
rs755680035 Chr.9: 20765006-20765006 on Build GRCh38
rs779092493 Chr.9: 20764949-20764949 on Build GRCh38
rs749938860 Chr.9: 20765077-20765077 on Build GRCh38
rs756700547 Chr.9: 20764931-20764931 on Build GRCh38
rs770911786 Chr.9: 20764960-20764960 on Build GRCh38
rs747145835 Chr.9: 20764958-20764958 on Build GRCh38
rs143683524 Chr.9: 20764972-20764972 on Build GRCh38
rs758744690 Chr.9: 20765018-20765018 on Build GRCh38
rs759618724 Chr.9: 20764966-20764966 on Build GRCh38
rs771055520 Chr.9: 20765030-20765030 on Build GRCh38
rs766074654 Chr.9: 20765091-20765091 on Build GRCh38
rs746134328 Chr.9: 20765040-20765040 on Build GRCh38
rs767080332 Chr.9: 20765074-20765074 on Build GRCh38
rs940174304 Chr.9: 20764941-20764941 on Build GRCh38
rs762811992 Chr.9: 20764973-20764973 on Build GRCh38
rs567596434 Chr.9: 20765102-20765102 on Build GRCh38
rs985521634 Chr.9: 20764982-20764982 on Build GRCh38
rs778319307 Chr.9: 20765023-20765023 on Build GRCh38
rs745506243 Chr.9: 20764940-20764940 on Build GRCh38
rs776916171 Chr.9: 20764965-20764965 on Build GRCh38
rs775833167 Chr.9: 20765048-20765048 on Build GRCh38
rs142943471 Chr.9: 20765064-20765064 on Build GRCh38
rs755626226 Chr.9: 20765087-20765087 on Build GRCh38
rs774438475 Chr.9: 20765056-20765056 on Build GRCh38
rs763980466 Chr.9: 20765055-20765055 on Build GRCh38
rs1007226199 Chr.9: 20765043-20765043 on Build GRCh38
rs748683454 Chr.9: 20764951-20764951 on Build GRCh38
rs200873740 Chr.9: 20764957-20764957 on Build GRCh38
rs748556974 Chr.9: 20765009-20765009 on Build GRCh38
rs1048595513 Chr.9: 20764928-20764928 on Build GRCh38
rs766874477 Chr.9: 20764992-20764992 on Build GRCh38
rs776522349 Chr.9: 20765037-20765037 on Build GRCh38
rs138365184 Chr.9: 20765046-20765046 on Build GRCh38
rs756861264 Chr.9: 20764989-20764989 on Build GRCh38
rs764176452 Chr.9: 20764981-20764981 on Build GRCh38
rs148930782 Chr.9: 20764929-20764929 on Build GRCh38
rs369821901 Chr.9: 20764996-20764996 on Build GRCh38

View Full Product Details