Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161049171-161049396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP30
Entrez Gene ID
257106
Gene Name
Rho GTPase activating protein 30
Gene Aliases
-
UniGene
Hs.389374
Gene Chromosome Location
Chr.1: 161046942-161069971 on Build GRCh38
Transcripts
NM_001025598.1 NM_001287600.1 NM_001287602.1 NM_181720.2 XM_005245070.2 XM_005245073.3 XM_011509391.2 XM_017000960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7079296 c.1778C>T p.S593F Chr.1: 161049243-161049243 on Build GRCh38
898437 c.1825T>G p.F609V Chr.1: 161049196-161049196 on Build GRCh38
3984397 c.1766G>A p.C589Y Chr.1: 161049255-161049255 on Build GRCh38
5704982 c.1709C>T p.P570L Chr.1: 161049312-161049312 on Build GRCh38
6450747 c.1747G>T p.V583F Chr.1: 161049274-161049274 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769634617 Chr.1: 161049199-161049199 on Build GRCh38
rs766759828 Chr.1: 161049337-161049337 on Build GRCh38
rs781318175 Chr.1: 161049248-161049248 on Build GRCh38
rs775108977 Chr.1: 161049286-161049286 on Build GRCh38
rs758788235 Chr.1: 161049238-161049238 on Build GRCh38
rs756135430 Chr.1: 161049378-161049378 on Build GRCh38
rs760223473 Chr.1: 161049308-161049308 on Build GRCh38
rs767053721 Chr.1: 161049217-161049217 on Build GRCh38
rs772832587 Chr.1: 161049222-161049222 on Build GRCh38
rs762914478 Chr.1: 161049205-161049205 on Build GRCh38
rs139973145 Chr.1: 161049267-161049267 on Build GRCh38
rs755488811 Chr.1: 161049345-161049345 on Build GRCh38
rs375000593 Chr.1: 161049369-161049369 on Build GRCh38
rs745808932 Chr.1: 161049261-161049261 on Build GRCh38
rs139339322 Chr.1: 161049209-161049209 on Build GRCh38
rs774223470 Chr.1: 161049255-161049255 on Build GRCh38
rs199749953 Chr.1: 161049334-161049334 on Build GRCh38
rs113542444 Chr.1: 161049237-161049237 on Build GRCh38
rs772503565 Chr.1: 161049202-161049202 on Build GRCh38
rs374832197 Chr.1: 161049218-161049218 on Build GRCh38
rs750851006 Chr.1: 161049232-161049232 on Build GRCh38
rs17854840 Chr.1: 161049251-161049251 on Build GRCh38
rs918030160 Chr.1: 161049274-161049274 on Build GRCh38
rs755079157 Chr.1: 161049243-161049243 on Build GRCh38
rs141806544 Chr.1: 161049329-161049329 on Build GRCh38
rs531763065 Chr.1: 161049200-161049200 on Build GRCh38
rs763863625 Chr.1: 161049317-161049317 on Build GRCh38
rs761202064 Chr.1: 161049241-161049241 on Build GRCh38
rs776016828 Chr.1: 161049211-161049211 on Build GRCh38
rs796212925 Chr.1: 161049312-161049312 on Build GRCh38
rs558271040 Chr.1: 161049352-161049352 on Build GRCh38
rs150907238 Chr.1: 161049242-161049242 on Build GRCh38
rs372930309 Chr.1: 161049296-161049296 on Build GRCh38
rs1006436567 Chr.1: 161049338-161049338 on Build GRCh38
rs3813609 Chr.1: 161049250-161049250 on Build GRCh38
rs149276784 Chr.1: 161049249-161049249 on Build GRCh38
rs372150791 Chr.1: 161049210-161049210 on Build GRCh38
rs548575480 Chr.1: 161049231-161049231 on Build GRCh38
rs752064064 Chr.1: 161049341-161049341 on Build GRCh38
rs767530546 Chr.1: 161049361-161049361 on Build GRCh38

View Full Product Details