Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 229526053-229526278 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB10
Entrez Gene ID
23456
Gene Name
ATP binding cassette subfamily B member 10
Gene Aliases
EST20237, M-ABC2, MTABC2
UniGene
Hs.17614
Gene Chromosome Location
Chr.1: 229516582-229558987 on Build GRCh38
Transcripts
NM_012089.2 XM_011544135.2 XM_011544136.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7122934 c.1756G>C p.E586Q Chr.1: 229526086-229526086 on Build GRCh38
905268 c.1748C>A p.S583Y Chr.1: 229526094-229526094 on Build GRCh38
354115 c.1778A>C p.D593A Chr.1: 229526064-229526064 on Build GRCh38
1320405 c.1750A>G p.I584V Chr.1: 229526092-229526092 on Build GRCh38
5797796 c.1768T>C p.Y590H Chr.1: 229526074-229526074 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746768406 Chr.1: 229526084-229526084 on Build GRCh38
rs781086132 Chr.1: 229526100-229526100 on Build GRCh38
rs113298661 Chr.1: 229526159-229526159 on Build GRCh38
rs375038821 Chr.1: 229526123-229526123 on Build GRCh38
rs919448289 Chr.1: 229526156-229526156 on Build GRCh38
rs1049147455 Chr.1: 229526180-229526180 on Build GRCh38
rs1039073320 Chr.1: 229526214-229526214 on Build GRCh38
rs772610116 Chr.1: 229526164-229526164 on Build GRCh38
rs773274959 Chr.1: 229526140-229526140 on Build GRCh38
rs771487964 Chr.1: 229526120-229526120 on Build GRCh38
rs756386808 Chr.1: 229526153-229526153 on Build GRCh38
rs564156405 Chr.1: 229526136-229526136 on Build GRCh38
rs74705869 Chr.1: 229526237-229526237 on Build GRCh38
rs370293229 Chr.1: 229526143-229526143 on Build GRCh38
rs774394369 Chr.1: 229526132-229526136 on Build GRCh38
rs901749770 Chr.1: 229526224-229526224 on Build GRCh38
rs988677175 Chr.1: 229526148-229526148 on Build GRCh38
rs776123169 Chr.1: 229526167-229526167 on Build GRCh38
rs943366750 Chr.1: 229526200-229526200 on Build GRCh38
rs754809765 Chr.1: 229526092-229526092 on Build GRCh38
rs779598408 Chr.1: 229526080-229526080 on Build GRCh38
rs147036546 Chr.1: 229526191-229526191 on Build GRCh38
rs1035780018 Chr.1: 229526118-229526118 on Build GRCh38

View Full Product Details