Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14401378-14401629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRE5
Entrez Gene ID
976
Gene Name
adhesion G protein-coupled receptor E5
Gene Aliases
CD97, TM7LN1
UniGene
Hs.466039
Gene Chromosome Location
Chr.19: 14381144-14408725 on Build GRCh38
Transcripts
NM_001025160.2 NM_001784.4 NM_078481.3 XM_011528451.2 XM_017027547.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6623969 c.1052C>T p.T351I Chr.19: 14401540-14401540 on Build GRCh38
3692249 c.1021C>A p.L341M Chr.19: 14401509-14401509 on Build GRCh38
5520355 c.960A>T p.P320P Chr.19: 14401448-14401448 on Build GRCh38
1304089 c.1045C>T p.P349S Chr.19: 14401533-14401533 on Build GRCh38
4917384 c.1005A>G p.E335E Chr.19: 14401493-14401493 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746207246 Chr.19: 14401479-14401479 on Build GRCh38
rs765247258 Chr.19: 14401535-14401535 on Build GRCh38
rs140629853 Chr.19: 14401412-14401412 on Build GRCh38
rs1003158202 Chr.19: 14401489-14401489 on Build GRCh38
rs770383003 Chr.19: 14401490-14401490 on Build GRCh38
rs770014400 Chr.19: 14401598-14401598 on Build GRCh38
rs775811992 Chr.19: 14401494-14401494 on Build GRCh38
rs767028684 Chr.19: 14401585-14401585 on Build GRCh38
rs939778146 Chr.19: 14401428-14401428 on Build GRCh38
rs761208416 Chr.19: 14401414-14401414 on Build GRCh38
rs376677729 Chr.19: 14401531-14401531 on Build GRCh38
rs956075882 Chr.19: 14401517-14401517 on Build GRCh38
rs367986615 Chr.19: 14401603-14401603 on Build GRCh38
rs750006287 Chr.19: 14401434-14401434 on Build GRCh38
rs1023067985 Chr.19: 14401573-14401573 on Build GRCh38
rs759273933 Chr.19: 14401534-14401534 on Build GRCh38
rs1012979522 Chr.19: 14401569-14401569 on Build GRCh38
rs747173884 Chr.19: 14401464-14401464 on Build GRCh38
rs767221670 Chr.19: 14401430-14401430 on Build GRCh38
rs771266840 Chr.19: 14401467-14401467 on Build GRCh38
rs758365972 Chr.19: 14401455-14401455 on Build GRCh38
rs1031506181 Chr.19: 14401499-14401499 on Build GRCh38
rs761418785 Chr.19: 14401496-14401496 on Build GRCh38
rs746258131 Chr.19: 14401596-14401596 on Build GRCh38
rs139618356 Chr.19: 14401539-14401539 on Build GRCh38
rs777230363 Chr.19: 14401588-14401589 on Build GRCh38
rs201125993 Chr.19: 14401582-14401582 on Build GRCh38
rs1043275920 Chr.19: 14401510-14401510 on Build GRCh38
rs199620684 Chr.19: 14401470-14401470 on Build GRCh38
rs758516153 Chr.19: 14401541-14401541 on Build GRCh38
rs772694731 Chr.19: 14401511-14401511 on Build GRCh38
rs778693810 Chr.19: 14401452-14401452 on Build GRCh38
rs778057042 Chr.19: 14401456-14401456 on Build GRCh38
rs61733714 Chr.19: 14401444-14401444 on Build GRCh38
rs559079747 Chr.19: 14401519-14401519 on Build GRCh38
rs760469641 Chr.19: 14401435-14401435 on Build GRCh38
rs751688182 Chr.19: 14401556-14401556 on Build GRCh38
rs370979328 Chr.19: 14401555-14401555 on Build GRCh38
rs757596890 Chr.19: 14401577-14401577 on Build GRCh38
rs753623982 Chr.19: 14401445-14401445 on Build GRCh38
rs781734469 Chr.19: 14401478-14401478 on Build GRCh38
rs537262542 Chr.19: 14401498-14401498 on Build GRCh38
rs923554059 Chr.19: 14401580-14401580 on Build GRCh38
rs16979636 Chr.19: 14401448-14401448 on Build GRCh38
rs375567054 Chr.19: 14401520-14401520 on Build GRCh38
rs765839682 Chr.19: 14401407-14401407 on Build GRCh38
rs752841394 Chr.19: 14401454-14401454 on Build GRCh38

View Full Product Details