Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 635278-635539 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL26
Entrez Gene ID
84326
Gene Name
methyltransferase like 26
Gene Aliases
C16orf13, JFP2
UniGene
Hs.239500
Gene Chromosome Location
Chr.16: 634427-636366 on Build GRCh38
Transcripts
NM_001040160.2 NM_001040161.2 NM_001040162.2 NM_001040165.2 NM_001288710.1 NM_032366.4 NR_109976.1 NR_109977.1 NR_109978.1 NR_109979.1 XM_011522713.2 XM_011522714.2

SNP Detail

SNP ID Chromosome Location
rs781371934 Chr.16: 635383-635384 on Build GRCh38
rs746520113 Chr.16: 635387-635387 on Build GRCh38
rs987742951 Chr.16: 635477-635477 on Build GRCh38
rs762269963 Chr.16: 635314-635314 on Build GRCh38
rs777508579 Chr.16: 635383-635383 on Build GRCh38
rs571456533 Chr.16: 635499-635499 on Build GRCh38
rs201877690 Chr.16: 635313-635313 on Build GRCh38
rs1015822895 Chr.16: 635479-635479 on Build GRCh38
rs369639213 Chr.16: 635361-635361 on Build GRCh38
rs200240022 Chr.16: 635332-635332 on Build GRCh38
rs376583737 Chr.16: 635379-635379 on Build GRCh38
rs997215226 Chr.16: 635444-635444 on Build GRCh38
rs749678888 Chr.16: 635300-635300 on Build GRCh38
rs567693046 Chr.16: 635509-635509 on Build GRCh38
rs913582528 Chr.16: 635486-635486 on Build GRCh38
rs1039092327 Chr.16: 635436-635436 on Build GRCh38
rs914490384 Chr.16: 635430-635430 on Build GRCh38
rs759403807 Chr.16: 635322-635322 on Build GRCh38
rs1051503015 Chr.16: 635450-635450 on Build GRCh38
rs1027109094 Chr.16: 635340-635340 on Build GRCh38
rs947051838 Chr.16: 635431-635431 on Build GRCh38
rs772923286 Chr.16: 635311-635311 on Build GRCh38
rs376115777 Chr.16: 635441-635441 on Build GRCh38
rs373126641 Chr.16: 635358-635358 on Build GRCh38
rs201138760 Chr.16: 635303-635303 on Build GRCh38
rs4331337 Chr.16: 635360-635360 on Build GRCh38
rs960547582 Chr.16: 635382-635382 on Build GRCh38
rs993110067 Chr.16: 635395-635395 on Build GRCh38
rs755219663 Chr.16: 635376-635376 on Build GRCh38
rs138177114 Chr.16: 635419-635420 on Build GRCh38
rs770304920 Chr.16: 635470-635470 on Build GRCh38
rs372232854 Chr.16: 635309-635309 on Build GRCh38
rs1023377592 Chr.16: 635505-635505 on Build GRCh38
rs373273169 Chr.16: 635376-635376 on Build GRCh38
rs751406664 Chr.16: 635375-635376 on Build GRCh38
rs778659791 Chr.16: 635301-635301 on Build GRCh38
rs755393036 Chr.16: 635372-635372 on Build GRCh38
rs776363307 Chr.16: 635316-635316 on Build GRCh38
rs767725491 Chr.16: 635353-635353 on Build GRCh38
rs891259078 Chr.16: 635451-635451 on Build GRCh38
rs963269628 Chr.16: 635487-635487 on Build GRCh38
rs748554507 Chr.16: 635391-635405 on Build GRCh38
rs1052992048 Chr.16: 635429-635429 on Build GRCh38
rs570558792 Chr.16: 635428-635428 on Build GRCh38
rs879134539 Chr.16: 635419-635419 on Build GRCh38
rs201064578 Chr.16: 635339-635339 on Build GRCh38
rs764888375 Chr.16: 635325-635325 on Build GRCh38

View Full Product Details