Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 5652932-5653161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB13
Entrez Gene ID
79754
Gene Name
ankyrin repeat and SOCS box containing 13
Gene Aliases
-
UniGene
Hs.445899
Gene Chromosome Location
Chr.10: 5638857-5666595 on Build GRCh38
Transcripts
NM_024701.3 NR_024581.1 NR_037164.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7317482 c.120G>C p.E40D Chr.10: 5652974-5652974 on Build GRCh38
4503062 c.62C>T p.T21I Chr.10: 5653032-5653032 on Build GRCh38
6428709 c.59G>T p.R20L Chr.10: 5653035-5653035 on Build GRCh38
7356555 c.133G>C p.V45L Chr.10: 5652961-5652961 on Build GRCh38
403618 c.93G>C p.E31D Chr.10: 5653001-5653001 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768135635 Chr.10: 5653079-5653079 on Build GRCh38
rs764333133 Chr.10: 5653042-5653042 on Build GRCh38
rs868270378 Chr.10: 5653008-5653008 on Build GRCh38
rs200257168 Chr.10: 5652971-5652971 on Build GRCh38
rs149399272 Chr.10: 5653055-5653055 on Build GRCh38
rs954648306 Chr.10: 5653036-5653036 on Build GRCh38
rs781109308 Chr.10: 5652952-5652952 on Build GRCh38
rs370196470 Chr.10: 5652966-5652966 on Build GRCh38
rs749029951 Chr.10: 5652967-5652967 on Build GRCh38
rs776279927 Chr.10: 5653029-5653029 on Build GRCh38
rs371031325 Chr.10: 5653011-5653011 on Build GRCh38
rs60413325 Chr.10: 5652961-5652961 on Build GRCh38
rs1020986539 Chr.10: 5653026-5653026 on Build GRCh38
rs940481152 Chr.10: 5653112-5653112 on Build GRCh38
rs562317874 Chr.10: 5653114-5653114 on Build GRCh38
rs899976187 Chr.10: 5653132-5653132 on Build GRCh38
rs557916184 Chr.10: 5653052-5653052 on Build GRCh38
rs147576380 Chr.10: 5653027-5653027 on Build GRCh38
rs575333736 Chr.10: 5653021-5653021 on Build GRCh38
rs763255307 Chr.10: 5653035-5653035 on Build GRCh38
rs772098164 Chr.10: 5653009-5653009 on Build GRCh38
rs142082937 Chr.10: 5653028-5653028 on Build GRCh38
rs568303793 Chr.10: 5652962-5652962 on Build GRCh38
rs879877788 Chr.10: 5652968-5652968 on Build GRCh38
rs868530624 Chr.10: 5652963-5652963 on Build GRCh38
rs757487246 Chr.10: 5653113-5653113 on Build GRCh38
rs780424485 Chr.10: 5652964-5652964 on Build GRCh38
rs556929057 Chr.10: 5653000-5653000 on Build GRCh38
rs79548623 Chr.10: 5653065-5653065 on Build GRCh38
rs773041877 Chr.10: 5653016-5653016 on Build GRCh38

View Full Product Details