Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 56585702-56585975 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM260
Entrez Gene ID
54916
Gene Name
transmembrane protein 260
Gene Aliases
C14orf101
UniGene
Hs.497253
Gene Chromosome Location
Chr.14: 56579791-56660629 on Build GRCh38
Transcripts
NM_017799.3 XM_005267771.1 XM_006720176.1 XM_006720178.1 XM_011536851.1 XM_011536852.1 XM_011536853.1 XM_017021379.1 XM_017021380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4051387 c.303T>C p.F101F Chr.14: 56585871-56585871 on Build GRCh38
165991 c.237_248del12 p.A81_L84delAITL Chr.14: 56585805-56585816 on Build GRCh38
271133 c.280G>A p.V94I Chr.14: 56585848-56585848 on Build GRCh38
698307 c.293G>T p.C98F Chr.14: 56585861-56585861 on Build GRCh38
3999297 c.225G>T p.T75T Chr.14: 56585793-56585793 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376818032 Chr.14: 56585852-56585852 on Build GRCh38
rs764958462 Chr.14: 56585794-56585794 on Build GRCh38
rs745620603 Chr.14: 56585836-56585836 on Build GRCh38
rs776331847 Chr.14: 56585795-56585796 on Build GRCh38
rs147933213 Chr.14: 56585889-56585889 on Build GRCh38
rs752685553 Chr.14: 56585746-56585746 on Build GRCh38
rs777174890 Chr.14: 56585927-56585927 on Build GRCh38
rs142968776 Chr.14: 56585824-56585824 on Build GRCh38
rs754899603 Chr.14: 56585910-56585910 on Build GRCh38
rs901409715 Chr.14: 56585736-56585736 on Build GRCh38
rs779263904 Chr.14: 56585834-56585834 on Build GRCh38
rs1016650061 Chr.14: 56585783-56585783 on Build GRCh38
rs777897605 Chr.14: 56585734-56585735 on Build GRCh38
rs764754867 Chr.14: 56585883-56585883 on Build GRCh38
rs749480739 Chr.14: 56585760-56585760 on Build GRCh38
rs781145315 Chr.14: 56585911-56585911 on Build GRCh38
rs759981900 Chr.14: 56585805-56585805 on Build GRCh38
rs775539519 Chr.14: 56585774-56585774 on Build GRCh38
rs771901254 Chr.14: 56585773-56585773 on Build GRCh38
rs200822110 Chr.14: 56585849-56585849 on Build GRCh38
rs757399392 Chr.14: 56585892-56585892 on Build GRCh38
rs61732665 Chr.14: 56585829-56585829 on Build GRCh38
rs761504259 Chr.14: 56585821-56585821 on Build GRCh38
rs767884944 Chr.14: 56585812-56585812 on Build GRCh38
rs778512344 Chr.14: 56585770-56585770 on Build GRCh38
rs986078641 Chr.14: 56585902-56585902 on Build GRCh38
rs3737171 Chr.14: 56585793-56585793 on Build GRCh38
rs201468977 Chr.14: 56585846-56585846 on Build GRCh38
rs772072020 Chr.14: 56585793-56585794 on Build GRCh38
rs765356854 Chr.14: 56585900-56585900 on Build GRCh38
rs750611315 Chr.14: 56585901-56585901 on Build GRCh38
rs774572764 Chr.14: 56585801-56585801 on Build GRCh38
rs144791089 Chr.14: 56585872-56585872 on Build GRCh38
rs371332026 Chr.14: 56585742-56585742 on Build GRCh38
rs772774176 Chr.14: 56585893-56585894 on Build GRCh38
rs768110235 Chr.14: 56585788-56585788 on Build GRCh38
rs757556156 Chr.14: 56585768-56585768 on Build GRCh38
rs188596516 Chr.14: 56585823-56585823 on Build GRCh38
rs896007543 Chr.14: 56585762-56585762 on Build GRCh38
rs753195195 Chr.14: 56585817-56585817 on Build GRCh38
rs749450115 Chr.14: 56585769-56585771 on Build GRCh38
rs761205145 Chr.14: 56585877-56585877 on Build GRCh38
rs767880042 Chr.14: 56585867-56585867 on Build GRCh38
rs762454717 Chr.14: 56585930-56585935 on Build GRCh38
rs528109756 Chr.14: 56585792-56585792 on Build GRCh38
rs540652503 Chr.14: 56585905-56585905 on Build GRCh38
rs756720473 Chr.14: 56585818-56585818 on Build GRCh38
rs747632686 Chr.14: 56585912-56585912 on Build GRCh38
rs764853765 Chr.14: 56585860-56585861 on Build GRCh38
rs140895546 Chr.14: 56585847-56585847 on Build GRCh38
rs371150228 Chr.14: 56585947-56585947 on Build GRCh38
rs570522479 Chr.14: 56585778-56585778 on Build GRCh38
rs376901455 Chr.14: 56585757-56585757 on Build GRCh38
rs751678026 Chr.14: 56585733-56585735 on Build GRCh38
rs144727961 Chr.14: 56585845-56585845 on Build GRCh38
rs552305187 Chr.14: 56585756-56585756 on Build GRCh38
rs762531305 Chr.14: 56585854-56585854 on Build GRCh38
rs769294895 Chr.14: 56585917-56585917 on Build GRCh38
rs529373773 Chr.14: 56585929-56585929 on Build GRCh38
rs746230777 Chr.14: 56585844-56585844 on Build GRCh38
rs746752753 Chr.14: 56585794-56585795 on Build GRCh38
rs758699644 Chr.14: 56585904-56585904 on Build GRCh38
rs922767038 Chr.14: 56585924-56585924 on Build GRCh38
rs747716407 Chr.14: 56585848-56585848 on Build GRCh38

View Full Product Details