Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53308735-53308970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAAS
Entrez Gene ID
8086
Gene Name
aladin WD repeat nucleoporin
Gene Aliases
AAA, AAASb, ADRACALA, ADRACALIN, ALADIN, GL003
UniGene
Hs.369144
Gene Chromosome Location
Chr.12: 53307456-53321628 on Build GRCh38
Transcripts
NM_001173466.1 NM_015665.5 XM_011538778.1 XM_011538780.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1932889 c.1025G>A p.R342Q Chr.12: 53308787-53308787 on Build GRCh38
1932890 c.1015G>C p.D339H Chr.12: 53308797-53308797 on Build GRCh38
940974 c.989G>A p.R330H Chr.12: 53308967-53308967 on Build GRCh38
3871943 c.1035C>T p.F345F Chr.12: 53308777-53308777 on Build GRCh38
3462649 c.1051C>T p.P351S Chr.12: 53308761-53308761 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922985890 Chr.12: 53308839-53308839 on Build GRCh38
rs753037128 Chr.12: 53308912-53308912 on Build GRCh38
rs1036729906 Chr.12: 53308899-53308899 on Build GRCh38
rs375457910 Chr.12: 53308857-53308857 on Build GRCh38
rs774292706 Chr.12: 53308834-53308834 on Build GRCh38
rs371901955 Chr.12: 53308777-53308777 on Build GRCh38
rs760456385 Chr.12: 53308845-53308845 on Build GRCh38
rs980119995 Chr.12: 53308782-53308782 on Build GRCh38
rs757303233 Chr.12: 53308788-53308788 on Build GRCh38
rs779774298 Chr.12: 53308849-53308849 on Build GRCh38
rs748827317 Chr.12: 53308863-53308863 on Build GRCh38
rs200312077 Chr.12: 53308948-53308948 on Build GRCh38
rs769989143 Chr.12: 53308803-53308803 on Build GRCh38
rs1039325300 Chr.12: 53308907-53308907 on Build GRCh38
rs909068099 Chr.12: 53308871-53308871 on Build GRCh38
rs755685527 Chr.12: 53308940-53308941 on Build GRCh38
rs909257867 Chr.12: 53308786-53308786 on Build GRCh38
rs779869524 Chr.12: 53308807-53308807 on Build GRCh38
rs368577869 Chr.12: 53308836-53308836 on Build GRCh38
rs764523065 Chr.12: 53308919-53308919 on Build GRCh38
rs973991055 Chr.12: 53308831-53308831 on Build GRCh38
rs956411998 Chr.12: 53308816-53308816 on Build GRCh38
rs375910859 Chr.12: 53308854-53308854 on Build GRCh38
rs768530319 Chr.12: 53308870-53308870 on Build GRCh38
rs753091732 Chr.12: 53308761-53308761 on Build GRCh38
rs140920186 Chr.12: 53308797-53308797 on Build GRCh38
rs374427447 Chr.12: 53308826-53308826 on Build GRCh38
rs749076938 Chr.12: 53308823-53308823 on Build GRCh38
rs746153303 Chr.12: 53308798-53308798 on Build GRCh38
rs1011231660 Chr.12: 53308888-53308888 on Build GRCh38
rs111986621 Chr.12: 53308940-53308940 on Build GRCh38
rs79184297 Chr.12: 53308879-53308879 on Build GRCh38
rs968298435 Chr.12: 53308773-53308773 on Build GRCh38
rs572186263 Chr.12: 53308787-53308787 on Build GRCh38
rs900728531 Chr.12: 53308917-53308917 on Build GRCh38
rs371424478 Chr.12: 53308916-53308916 on Build GRCh38
rs200674403 Chr.12: 53308934-53308934 on Build GRCh38
rs1053277248 Chr.12: 53308875-53308875 on Build GRCh38
rs113635763 Chr.12: 53308840-53308840 on Build GRCh38

View Full Product Details