Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66567265-66567522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSF
Entrez Gene ID
8722
Gene Name
cathepsin F
Gene Aliases
CATSF, CLN13
UniGene
Hs.11590
Gene Chromosome Location
Chr.11: 66563463-66568576 on Build GRCh38
Transcripts
NM_003793.3 XM_011545328.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3398063 c.538C>T p.P180S Chr.11: 66567315-66567315 on Build GRCh38
2040474 c.490A>G p.S164G Chr.11: 66567485-66567485 on Build GRCh38
7099431 c.469G>A p.D157N Chr.11: 66567506-66567506 on Build GRCh38
127744 c.573C>T p.V191V Chr.11: 66567280-66567280 on Build GRCh38
4035754 c.549G>A p.M183I Chr.11: 66567304-66567304 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548621329 Chr.11: 66567364-66567364 on Build GRCh38
rs150926626 Chr.11: 66567441-66567441 on Build GRCh38
rs773296151 Chr.11: 66567498-66567498 on Build GRCh38
rs777872865 Chr.11: 66567329-66567329 on Build GRCh38
rs773279173 Chr.11: 66567418-66567418 on Build GRCh38
rs751783877 Chr.11: 66567407-66567407 on Build GRCh38
rs780652380 Chr.11: 66567484-66567484 on Build GRCh38
rs528679828 Chr.11: 66567307-66567307 on Build GRCh38
rs369416976 Chr.11: 66567483-66567483 on Build GRCh38
rs772366380 Chr.11: 66567497-66567497 on Build GRCh38
rs756575937 Chr.11: 66567369-66567369 on Build GRCh38
rs765760543 Chr.11: 66567311-66567312 on Build GRCh38
rs756862766 Chr.11: 66567450-66567450 on Build GRCh38
rs751230557 Chr.11: 66567442-66567442 on Build GRCh38
rs755417013 Chr.11: 66567490-66567490 on Build GRCh38
rs748390300 Chr.11: 66567495-66567495 on Build GRCh38
rs371296361 Chr.11: 66567366-66567366 on Build GRCh38
rs749889136 Chr.11: 66567462-66567462 on Build GRCh38
rs757428276 Chr.11: 66567429-66567430 on Build GRCh38
rs758444193 Chr.11: 66567376-66567376 on Build GRCh38
rs945569732 Chr.11: 66567345-66567345 on Build GRCh38
rs914070454 Chr.11: 66567337-66567337 on Build GRCh38
rs762774640 Chr.11: 66567436-66567436 on Build GRCh38
rs900704359 Chr.11: 66567447-66567447 on Build GRCh38
rs753741916 Chr.11: 66567297-66567297 on Build GRCh38
rs751179004 Chr.11: 66567360-66567360 on Build GRCh38
rs764382629 Chr.11: 66567401-66567402 on Build GRCh38
rs765279763 Chr.11: 66567428-66567428 on Build GRCh38
rs757481259 Chr.11: 66567422-66567422 on Build GRCh38
rs781322870 Chr.11: 66567439-66567439 on Build GRCh38
rs749730056 Chr.11: 66567373-66567373 on Build GRCh38
rs763377999 Chr.11: 66567395-66567410 on Build GRCh38
rs562176915 Chr.11: 66567383-66567383 on Build GRCh38
rs200050648 Chr.11: 66567398-66567398 on Build GRCh38
rs752481810 Chr.11: 66567340-66567340 on Build GRCh38
rs899212002 Chr.11: 66567471-66567471 on Build GRCh38
rs374291363 Chr.11: 66567397-66567397 on Build GRCh38
rs749792353 Chr.11: 66567486-66567486 on Build GRCh38
rs748331186 Chr.11: 66567404-66567404 on Build GRCh38
rs775282716 Chr.11: 66567434-66567434 on Build GRCh38
rs527638704 Chr.11: 66567402-66567417 on Build GRCh38
rs780293872 Chr.11: 66567371-66567371 on Build GRCh38
rs766940123 Chr.11: 66567453-66567453 on Build GRCh38
rs759408416 Chr.11: 66567427-66567427 on Build GRCh38
rs763799576 Chr.11: 66567358-66567358 on Build GRCh38
rs372789274 Chr.11: 66567413-66567413 on Build GRCh38
rs781286634 Chr.11: 66567450-66567451 on Build GRCh38
rs753069903 Chr.11: 66567348-66567348 on Build GRCh38
rs150481606 Chr.11: 66567316-66567316 on Build GRCh38
rs753203173 Chr.11: 66567303-66567303 on Build GRCh38

View Full Product Details