Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154432397-154432644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6AP1
Entrez Gene ID
537
Gene Name
ATPase H+ transporting accessory protein 1
Gene Aliases
16A, ATP6IP1, ATP6S1, Ac45, CF2, VATPS1, XAP-3, XAP3
UniGene
Hs.633817
Gene Chromosome Location
Chr.X: 154428632-154436517 on Build GRCh38
Transcripts
NM_001183.5 XM_011531179.1

SNP Detail

SNP ID Chromosome Location
rs200084311 Chr.X: 154432480-154432480 on Build GRCh38
rs140841742 Chr.X: 154432441-154432441 on Build GRCh38
rs782667594 Chr.X: 154432474-154432474 on Build GRCh38
rs781961520 Chr.X: 154432597-154432597 on Build GRCh38
rs782606249 Chr.X: 154432454-154432454 on Build GRCh38
rs782325652 Chr.X: 154432469-154432469 on Build GRCh38
rs782177963 Chr.X: 154432617-154432617 on Build GRCh38
rs782075809 Chr.X: 154432484-154432484 on Build GRCh38
rs782179986 Chr.X: 154432468-154432468 on Build GRCh38
rs782288870 Chr.X: 154432476-154432476 on Build GRCh38
rs782537350 Chr.X: 154432442-154432442 on Build GRCh38
rs781925757 Chr.X: 154432491-154432491 on Build GRCh38
rs374882679 Chr.X: 154432451-154432451 on Build GRCh38
rs909684160 Chr.X: 154432544-154432544 on Build GRCh38
rs941468530 Chr.X: 154432611-154432611 on Build GRCh38
rs782724094 Chr.X: 154432490-154432490 on Build GRCh38
rs200943680 Chr.X: 154432440-154432440 on Build GRCh38
rs782677279 Chr.X: 154432465-154432465 on Build GRCh38
rs144602046 Chr.X: 154432450-154432450 on Build GRCh38
rs782690734 Chr.X: 154432443-154432443 on Build GRCh38
rs781855202 Chr.X: 154432449-154432449 on Build GRCh38
rs182723602 Chr.X: 154432539-154432539 on Build GRCh38
rs929867343 Chr.X: 154432522-154432522 on Build GRCh38
rs148286758 Chr.X: 154432537-154432537 on Build GRCh38
rs781997075 Chr.X: 154432482-154432482 on Build GRCh38

View Full Product Details