Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38674166-38674405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf96
Entrez Gene ID
100170841
Gene Name
chromosome 17 open reading frame 96
Gene Aliases
PRR28
UniGene
Hs.445574
Gene Chromosome Location
Chr.17: 38671703-38674934 on Build GRCh38
Transcripts
NM_001130677.1

SNP Detail

SNP ID Chromosome Location
rs572143190 Chr.17: 38674282-38674282 on Build GRCh38
rs760205814 Chr.17: 38674244-38674244 on Build GRCh38
rs1029512819 Chr.17: 38674226-38674226 on Build GRCh38
rs541981015 Chr.17: 38674188-38674188 on Build GRCh38
rs992252693 Chr.17: 38674351-38674351 on Build GRCh38
rs563130483 Chr.17: 38674386-38674386 on Build GRCh38
rs184645580 Chr.17: 38674319-38674319 on Build GRCh38
rs541031175 Chr.17: 38674288-38674288 on Build GRCh38
rs79676758 Chr.17: 38674309-38674309 on Build GRCh38
rs1031580898 Chr.17: 38674183-38674183 on Build GRCh38
rs552572187 Chr.17: 38674382-38674382 on Build GRCh38
rs1056434444 Chr.17: 38674256-38674256 on Build GRCh38
rs985713433 Chr.17: 38674185-38674185 on Build GRCh38
rs766878152 Chr.17: 38674245-38674245 on Build GRCh38
rs891518726 Chr.17: 38674354-38674354 on Build GRCh38
rs1017466055 Chr.17: 38674278-38674278 on Build GRCh38
rs753118395 Chr.17: 38674323-38674323 on Build GRCh38
rs895405475 Chr.17: 38674265-38674265 on Build GRCh38
rs758636860 Chr.17: 38674329-38674329 on Build GRCh38
rs939617129 Chr.17: 38674341-38674341 on Build GRCh38
rs778163827 Chr.17: 38674337-38674337 on Build GRCh38
rs111565436 Chr.17: 38674206-38674206 on Build GRCh38
rs948260088 Chr.17: 38674379-38674379 on Build GRCh38
rs1031429121 Chr.17: 38674342-38674342 on Build GRCh38
rs916757074 Chr.17: 38674360-38674360 on Build GRCh38
rs985557592 Chr.17: 38674271-38674271 on Build GRCh38
rs1037110048 Chr.17: 38674219-38674219 on Build GRCh38

View Full Product Details