Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27130700-27130939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD1
Entrez Gene ID
84696
Gene Name
abhydrolase domain containing 1
Gene Aliases
LABH1
UniGene
Hs.375791
Gene Chromosome Location
Chr.2: 27123780-27130812 on Build GRCh38
Transcripts
NM_032604.3 XM_011533135.2 XM_011533136.2 XM_011533138.2 NM_013388.4 XM_006711914.2 XM_011532471.2 XM_011532472.2

Gene Symbol
PREB
Entrez Gene ID
10113
Gene Name
prolactin regulatory element binding
Gene Aliases
SEC12
UniGene
Hs.279784
Gene Chromosome Location
Chr.2: 27130756-27134674 on Build GRCh38
Transcripts
NM_032604.3 XM_011533135.2 XM_011533136.2 XM_011533138.2 NM_013388.4 XM_006711914.2 XM_011532471.2 XM_011532472.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1019524 c.1189C>A p.L397I Chr.2: 27130715-27130715 on Build GRCh38
5899378 c.1208G>A p.R403K Chr.2: 27130734-27130734 on Build GRCh38
7260785 c.1212C>T p.N404N Chr.2: 27130738-27130738 on Build GRCh38
5938814 c.1204G>A p.D402N Chr.2: 27130730-27130730 on Build GRCh38
5708175 c.1184G>A p.R395K Chr.2: 27130710-27130710 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922436033 Chr.2: 27130775-27130775 on Build GRCh38
rs1013130335 Chr.2: 27130912-27130912 on Build GRCh38
rs573617544 Chr.2: 27130773-27130773 on Build GRCh38
rs778332213 Chr.2: 27130776-27130776 on Build GRCh38
rs574063161 Chr.2: 27130750-27130750 on Build GRCh38
rs562319173 Chr.2: 27130870-27130870 on Build GRCh38
rs150770274 Chr.2: 27130744-27130744 on Build GRCh38
rs745623465 Chr.2: 27130795-27130795 on Build GRCh38
rs771966903 Chr.2: 27130779-27130779 on Build GRCh38
rs41288799 Chr.2: 27130762-27130762 on Build GRCh38
rs748904063 Chr.2: 27130771-27130771 on Build GRCh38
rs750613988 Chr.2: 27130721-27130721 on Build GRCh38
rs746812273 Chr.2: 27130792-27130792 on Build GRCh38
rs769501676 Chr.2: 27130812-27130812 on Build GRCh38
rs775153012 Chr.2: 27130851-27130851 on Build GRCh38
rs1024937580 Chr.2: 27130741-27130741 on Build GRCh38
rs775429009 Chr.2: 27130787-27130787 on Build GRCh38
rs933746527 Chr.2: 27130782-27130782 on Build GRCh38
rs540922561 Chr.2: 27130818-27130818 on Build GRCh38
rs746930675 Chr.2: 27130723-27130723 on Build GRCh38
rs543827411 Chr.2: 27130891-27130891 on Build GRCh38
rs756415500 Chr.2: 27130761-27130761 on Build GRCh38
rs763148217 Chr.2: 27130738-27130738 on Build GRCh38
rs753065089 Chr.2: 27130758-27130758 on Build GRCh38
rs755496103 Chr.2: 27130748-27130748 on Build GRCh38
rs745366093 Chr.2: 27130778-27130778 on Build GRCh38
rs781720939 Chr.2: 27130749-27130749 on Build GRCh38
rs763653700 Chr.2: 27130896-27130896 on Build GRCh38
rs992070158 Chr.2: 27130726-27130726 on Build GRCh38
rs186368898 Chr.2: 27130876-27130876 on Build GRCh38

View Full Product Details