Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 152974827-152975100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO5
Entrez Gene ID
26271
Gene Name
F-box protein 5
Gene Aliases
EMI1, FBX5, Fbxo31
UniGene
Hs.520506
Gene Chromosome Location
Chr.6: 152970523-152983605 on Build GRCh38
Transcripts
NM_001142522.1 NM_012177.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1169063 c.629A>T p.K210I Chr.6: 152975096-152975096 on Build GRCh38
6301745 c.657G>A p.M219I Chr.6: 152975068-152975068 on Build GRCh38
4684027 c.715delA p.M239fs*3 Chr.6: 152975010-152975010 on Build GRCh38
4748009 c.750_751delCT p.F251fs*17 Chr.6: 152974974-152974975 on Build GRCh38
3021548 c.650G>A p.R217Q Chr.6: 152975075-152975075 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564355898 Chr.6: 152974926-152974926 on Build GRCh38
rs780648667 Chr.6: 152974981-152974981 on Build GRCh38
rs772321512 Chr.6: 152975047-152975047 on Build GRCh38
rs773406316 Chr.6: 152975052-152975052 on Build GRCh38
rs532952813 Chr.6: 152974975-152974975 on Build GRCh38
rs138580404 Chr.6: 152974998-152974998 on Build GRCh38
rs755420488 Chr.6: 152974879-152974879 on Build GRCh38
rs767652352 Chr.6: 152974936-152974936 on Build GRCh38
rs375619264 Chr.6: 152974909-152974909 on Build GRCh38
rs201171632 Chr.6: 152974930-152974930 on Build GRCh38
rs893159365 Chr.6: 152974853-152974853 on Build GRCh38
rs774448542 Chr.6: 152975042-152975042 on Build GRCh38
rs746518936 Chr.6: 152974891-152974891 on Build GRCh38
rs745567094 Chr.6: 152974993-152974993 on Build GRCh38
rs779399211 Chr.6: 152974970-152974970 on Build GRCh38
rs770361783 Chr.6: 152974892-152974892 on Build GRCh38
rs766819016 Chr.6: 152974950-152974950 on Build GRCh38
rs756791089 Chr.6: 152974980-152974980 on Build GRCh38
rs769828399 Chr.6: 152974956-152974956 on Build GRCh38
rs1030948525 Chr.6: 152975011-152975011 on Build GRCh38
rs775523395 Chr.6: 152975043-152975043 on Build GRCh38
rs911716305 Chr.6: 152974896-152974896 on Build GRCh38
rs374443845 Chr.6: 152974966-152974966 on Build GRCh38
rs779964165 Chr.6: 152975008-152975008 on Build GRCh38
rs752512537 Chr.6: 152975010-152975010 on Build GRCh38
rs560092200 Chr.6: 152974851-152974851 on Build GRCh38
rs777401301 Chr.6: 152974888-152974888 on Build GRCh38
rs761018036 Chr.6: 152975058-152975058 on Build GRCh38
rs201579649 Chr.6: 152974945-152974945 on Build GRCh38
rs148657499 Chr.6: 152974917-152974917 on Build GRCh38
rs915518949 Chr.6: 152974977-152974977 on Build GRCh38
rs945613195 Chr.6: 152975033-152975033 on Build GRCh38
rs370209486 Chr.6: 152974924-152974924 on Build GRCh38
rs950613626 Chr.6: 152974884-152974884 on Build GRCh38
rs754142126 Chr.6: 152974955-152974955 on Build GRCh38
rs994807366 Chr.6: 152974868-152974868 on Build GRCh38
rs766507735 Chr.6: 152975066-152975066 on Build GRCh38
rs749037838 Chr.6: 152975014-152975014 on Build GRCh38
rs763003006 Chr.6: 152974918-152974918 on Build GRCh38
rs780836670 Chr.6: 152974895-152974895 on Build GRCh38
rs750710095 Chr.6: 152974941-152974941 on Build GRCh38
rs1028020113 Chr.6: 152974874-152974874 on Build GRCh38
rs1011506358 Chr.6: 152974870-152974870 on Build GRCh38
rs768436308 Chr.6: 152975023-152975023 on Build GRCh38

View Full Product Details