Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 128946822-128947095 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRF5
Entrez Gene ID
3663
Gene Name
interferon regulatory factor 5
Gene Aliases
SLEB10
UniGene
Hs.521181
Gene Chromosome Location
Chr.7: 128937737-128950042 on Build GRCh38
Transcripts
NM_001098627.3 NM_001098629.2 NM_001098630.2 NM_001242452.2 NM_032643.4 XM_005250317.3 XM_006715974.2 XM_011516158.2 XM_011516159.2 XM_011516160.1 XM_011516161.1 XM_011516162.1 XM_011516163.2 XM_011516164.1

SNP Detail

SNP ID Chromosome Location
rs760237069 Chr.7: 128947060-128947060 on Build GRCh38
rs770852885 Chr.7: 128947007-128947007 on Build GRCh38
rs1048222480 Chr.7: 128946911-128946911 on Build GRCh38
rs535574945 Chr.7: 128946864-128946864 on Build GRCh38
rs777926297 Chr.7: 128946992-128946992 on Build GRCh38
rs763385119 Chr.7: 128947049-128947049 on Build GRCh38
rs375199151 Chr.7: 128947050-128947050 on Build GRCh38
rs527920344 Chr.7: 128946931-128946931 on Build GRCh38
rs746421090 Chr.7: 128947024-128947024 on Build GRCh38
rs200163710 Chr.7: 128947064-128947064 on Build GRCh38
rs758656823 Chr.7: 128946982-128946982 on Build GRCh38
rs866193368 Chr.7: 128946894-128946894 on Build GRCh38
rs1022425576 Chr.7: 128947052-128947052 on Build GRCh38
rs549413419 Chr.7: 128946875-128946875 on Build GRCh38
rs1037371666 Chr.7: 128946957-128946957 on Build GRCh38
rs747146617 Chr.7: 128946995-128946995 on Build GRCh38
rs1004378985 Chr.7: 128946848-128946848 on Build GRCh38
rs545707851 Chr.7: 128946863-128946863 on Build GRCh38
rs753371056 Chr.7: 128947065-128947065 on Build GRCh38
rs375259154 Chr.7: 128946882-128946882 on Build GRCh38
rs776039297 Chr.7: 128947044-128947044 on Build GRCh38
rs866877140 Chr.7: 128947003-128947003 on Build GRCh38
rs192829776 Chr.7: 128946981-128946981 on Build GRCh38
rs932438631 Chr.7: 128946874-128946874 on Build GRCh38
rs968644433 Chr.7: 128947005-128947005 on Build GRCh38
rs1004098914 Chr.7: 128946944-128946944 on Build GRCh38
rs772699436 Chr.7: 128947054-128947054 on Build GRCh38
rs770420647 Chr.7: 128947034-128947034 on Build GRCh38
rs781731127 Chr.7: 128947013-128947013 on Build GRCh38

View Full Product Details