Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38716793-38717066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTPBP1
Entrez Gene ID
9567
Gene Name
GTP binding protein 1
Gene Aliases
GP-1, GP1, HSPC018
UniGene
Hs.276925
Gene Chromosome Location
Chr.22: 38704561-38738265 on Build GRCh38
Transcripts
NM_004286.4 XM_011530537.2 XM_017029099.1 XM_017029100.1 XM_017029101.1 XM_017029102.1 XM_017029103.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5366492 c.692A>G p.H231R Chr.22: 38716858-38716858 on Build GRCh38
77825 c.670G>C p.V224L Chr.22: 38716836-38716836 on Build GRCh38
5057815 c.775T>C p.Y259H Chr.22: 38716941-38716941 on Build GRCh38
7245737 c.657C>T p.D219D Chr.22: 38716823-38716823 on Build GRCh38
3554417 c.807C>G p.G269G Chr.22: 38716973-38716973 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754343843 Chr.22: 38717040-38717040 on Build GRCh38
rs765222657 Chr.22: 38716863-38716863 on Build GRCh38
rs766471698 Chr.22: 38716892-38716892 on Build GRCh38
rs200575529 Chr.22: 38717016-38717016 on Build GRCh38
rs373810274 Chr.22: 38717039-38717039 on Build GRCh38
rs768486963 Chr.22: 38717009-38717009 on Build GRCh38
rs745400646 Chr.22: 38716826-38716826 on Build GRCh38
rs755252334 Chr.22: 38716901-38716901 on Build GRCh38
rs746644396 Chr.22: 38717008-38717008 on Build GRCh38
rs78490458 Chr.22: 38716991-38716991 on Build GRCh38
rs775302461 Chr.22: 38717022-38717028 on Build GRCh38
rs933638738 Chr.22: 38717017-38717017 on Build GRCh38
rs539803174 Chr.22: 38717007-38717007 on Build GRCh38
rs369739803 Chr.22: 38716961-38716961 on Build GRCh38
rs117911959 Chr.22: 38717025-38717025 on Build GRCh38
rs933220939 Chr.22: 38716847-38716847 on Build GRCh38
rs753013791 Chr.22: 38717031-38717031 on Build GRCh38
rs775244545 Chr.22: 38716835-38716835 on Build GRCh38
rs532123467 Chr.22: 38717014-38717014 on Build GRCh38
rs756536933 Chr.22: 38717032-38717032 on Build GRCh38
rs201300285 Chr.22: 38716829-38716829 on Build GRCh38
rs771584813 Chr.22: 38716992-38716992 on Build GRCh38
rs150400452 Chr.22: 38716859-38716859 on Build GRCh38
rs761834756 Chr.22: 38716862-38716862 on Build GRCh38
rs760440126 Chr.22: 38716841-38716841 on Build GRCh38
rs756523233 Chr.22: 38716919-38716919 on Build GRCh38
rs761680359 Chr.22: 38717013-38717013 on Build GRCh38
rs975136134 Chr.22: 38717003-38717003 on Build GRCh38
rs763114779 Chr.22: 38716879-38716879 on Build GRCh38
rs776623270 Chr.22: 38716860-38716860 on Build GRCh38
rs138114417 Chr.22: 38716916-38716916 on Build GRCh38
rs766469492 Chr.22: 38717022-38717022 on Build GRCh38
rs564641295 Chr.22: 38717011-38717011 on Build GRCh38
rs751758343 Chr.22: 38716900-38716900 on Build GRCh38
rs193190760 Chr.22: 38716874-38716874 on Build GRCh38
rs774481957 Chr.22: 38717023-38717023 on Build GRCh38
rs762847870 Chr.22: 38717020-38717020 on Build GRCh38

View Full Product Details