Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 49920081-49920328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRELD2
Entrez Gene ID
79174
Gene Name
cysteine rich with EGF like domains 2
Gene Aliases
-
UniGene
Hs.211282
Gene Chromosome Location
Chr.22: 49918630-49927540 on Build GRCh38
Transcripts
NM_001135101.2 NM_001284317.1 NM_001284318.1 NM_024324.4 NR_104295.1 XM_005261737.2 XM_005261738.4 XM_011530372.2 XM_017028938.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1035282 c.348C>T p.F116F Chr.22: 49920180-49920180 on Build GRCh38
4104973 c.408C>T p.D136D Chr.22: 49920240-49920240 on Build GRCh38
4104974 c.408C>T p.D136D Chr.22: 49920240-49920240 on Build GRCh38
7416627 c.340G>T p.D114Y Chr.22: 49920172-49920172 on Build GRCh38
7416626 c.340G>T p.D114Y Chr.22: 49920172-49920172 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764121862 Chr.22: 49920265-49920265 on Build GRCh38
rs11545764 Chr.22: 49920219-49920219 on Build GRCh38
rs183002505 Chr.22: 49920251-49920251 on Build GRCh38
rs761987095 Chr.22: 49920241-49920241 on Build GRCh38
rs770593818 Chr.22: 49920114-49920114 on Build GRCh38
rs763149806 Chr.22: 49920250-49920250 on Build GRCh38
rs772618402 Chr.22: 49920228-49920228 on Build GRCh38
rs756682085 Chr.22: 49920209-49920209 on Build GRCh38
rs941541269 Chr.22: 49920207-49920207 on Build GRCh38
rs755814898 Chr.22: 49920259-49920259 on Build GRCh38
rs918098843 Chr.22: 49920293-49920293 on Build GRCh38
rs199752594 Chr.22: 49920282-49920282 on Build GRCh38
rs143216272 Chr.22: 49920178-49920178 on Build GRCh38
rs761246139 Chr.22: 49920159-49920159 on Build GRCh38
rs756043231 Chr.22: 49920183-49920183 on Build GRCh38
rs756038625 Chr.22: 49920238-49920238 on Build GRCh38
rs114332575 Chr.22: 49920232-49920232 on Build GRCh38
rs752690863 Chr.22: 49920181-49920181 on Build GRCh38
rs1037578379 Chr.22: 49920246-49920246 on Build GRCh38
rs1045432018 Chr.22: 49920302-49920302 on Build GRCh38
rs200229316 Chr.22: 49920143-49920143 on Build GRCh38
rs764143058 Chr.22: 49920160-49920160 on Build GRCh38
rs897426126 Chr.22: 49920297-49920297 on Build GRCh38
rs767327101 Chr.22: 49920179-49920179 on Build GRCh38
rs11545765 Chr.22: 49920247-49920247 on Build GRCh38
rs372572017 Chr.22: 49920252-49920252 on Build GRCh38
rs950332644 Chr.22: 49920301-49920301 on Build GRCh38
rs780121954 Chr.22: 49920260-49920260 on Build GRCh38
rs371045417 Chr.22: 49920281-49920281 on Build GRCh38
rs746266395 Chr.22: 49920124-49920124 on Build GRCh38
rs780616604 Chr.22: 49920286-49920286 on Build GRCh38
rs781233113 Chr.22: 49920293-49920295 on Build GRCh38
rs765522989 Chr.22: 49920168-49920168 on Build GRCh38
rs747403079 Chr.22: 49920289-49920289 on Build GRCh38
rs551660195 Chr.22: 49920180-49920180 on Build GRCh38
rs773823948 Chr.22: 49920121-49920121 on Build GRCh38
rs778268944 Chr.22: 49920280-49920280 on Build GRCh38
rs745840668 Chr.22: 49920300-49920302 on Build GRCh38
rs79137702 Chr.22: 49920162-49920162 on Build GRCh38
rs111385776 Chr.22: 49920198-49920198 on Build GRCh38
rs555701929 Chr.22: 49920135-49920135 on Build GRCh38
rs776150017 Chr.22: 49920146-49920146 on Build GRCh38
rs966326268 Chr.22: 49920137-49920137 on Build GRCh38
rs369398193 Chr.22: 49920303-49920303 on Build GRCh38
rs768866055 Chr.22: 49920291-49920291 on Build GRCh38
rs761993336 Chr.22: 49920163-49920163 on Build GRCh38
rs753650046 Chr.22: 49920271-49920271 on Build GRCh38
rs376383650 Chr.22: 49920273-49920273 on Build GRCh38
rs748850650 Chr.22: 49920208-49920208 on Build GRCh38
rs745456519 Chr.22: 49920220-49920220 on Build GRCh38

View Full Product Details