Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 170084649-170084898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR160
Entrez Gene ID
26996
Gene Name
G protein-coupled receptor 160
Gene Aliases
GPCR1, GPCR150
UniGene
Hs.231320
Gene Chromosome Location
Chr.3: 170037323-170085415 on Build GRCh38
Transcripts
NM_014373.2 XM_005247346.3 XM_005247347.3 XM_006713582.3 XM_011512678.2 XM_017006161.1 XM_017006162.1 XM_017006163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5587904 c.920C>T p.P307L Chr.3: 170084892-170084892 on Build GRCh38
5814748 c.893A>G p.K298R Chr.3: 170084865-170084865 on Build GRCh38
4115147 c.906A>C p.K302N Chr.3: 170084878-170084878 on Build GRCh38
1693793 c.913G>A p.G305R Chr.3: 170084885-170084885 on Build GRCh38
4115146 c.868A>G p.T290A Chr.3: 170084840-170084840 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760706542 Chr.3: 170084830-170084830 on Build GRCh38
rs146297186 Chr.3: 170084813-170084813 on Build GRCh38
rs764007123 Chr.3: 170084770-170084770 on Build GRCh38
rs773645860 Chr.3: 170084707-170084710 on Build GRCh38
rs146985269 Chr.3: 170084790-170084790 on Build GRCh38
rs764782116 Chr.3: 170084840-170084840 on Build GRCh38
rs1054333575 Chr.3: 170084748-170084748 on Build GRCh38
rs764735904 Chr.3: 170084777-170084777 on Build GRCh38
rs552972161 Chr.3: 170084846-170084846 on Build GRCh38
rs775559956 Chr.3: 170084768-170084768 on Build GRCh38
rs757984039 Chr.3: 170084787-170084787 on Build GRCh38
rs771901447 Chr.3: 170084761-170084761 on Build GRCh38
rs142366644 Chr.3: 170084792-170084792 on Build GRCh38
rs139558562 Chr.3: 170084828-170084828 on Build GRCh38
rs913131256 Chr.3: 170084775-170084775 on Build GRCh38
rs757623733 Chr.3: 170084851-170084851 on Build GRCh38
rs748401347 Chr.3: 170084733-170084733 on Build GRCh38
rs544108325 Chr.3: 170084697-170084697 on Build GRCh38
rs768510633 Chr.3: 170084833-170084833 on Build GRCh38
rs771360982 Chr.3: 170084756-170084758 on Build GRCh38
rs758830676 Chr.3: 170084682-170084682 on Build GRCh38
rs773179424 Chr.3: 170084749-170084749 on Build GRCh38
rs751971278 Chr.3: 170084722-170084722 on Build GRCh38
rs781559786 Chr.3: 170084800-170084800 on Build GRCh38
rs747320666 Chr.3: 170084702-170084702 on Build GRCh38
rs148749842 Chr.3: 170084776-170084776 on Build GRCh38
rs748213865 Chr.3: 170084680-170084681 on Build GRCh38
rs199980016 Chr.3: 170084782-170084782 on Build GRCh38
rs1049269309 Chr.3: 170084760-170084760 on Build GRCh38
rs769892382 Chr.3: 170084688-170084688 on Build GRCh38
rs534725792 Chr.3: 170084819-170084819 on Build GRCh38
rs749894318 Chr.3: 170084780-170084780 on Build GRCh38
rs771096280 Chr.3: 170084824-170084824 on Build GRCh38
rs749121327 Chr.3: 170084698-170084698 on Build GRCh38
rs371588925 Chr.3: 170084812-170084812 on Build GRCh38
rs1001508653 Chr.3: 170084843-170084843 on Build GRCh38
rs762337181 Chr.3: 170084863-170084863 on Build GRCh38
rs760528134 Chr.3: 170084769-170084769 on Build GRCh38
rs749317743 Chr.3: 170084821-170084821 on Build GRCh38
rs761765017 Chr.3: 170084837-170084837 on Build GRCh38
rs759452986 Chr.3: 170084757-170084757 on Build GRCh38
rs756990605 Chr.3: 170084726-170084726 on Build GRCh38
rs769816175 Chr.3: 170084740-170084740 on Build GRCh38
rs750880925 Chr.3: 170084675-170084675 on Build GRCh38
rs769057032 Chr.3: 170084721-170084721 on Build GRCh38
rs377308505 Chr.3: 170084815-170084815 on Build GRCh38
rs763512509 Chr.3: 170084708-170084709 on Build GRCh38
rs781475418 Chr.3: 170084852-170084852 on Build GRCh38
rs776570089 Chr.3: 170084835-170084835 on Build GRCh38
rs752100174 Chr.3: 170084791-170084791 on Build GRCh38

View Full Product Details