Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27630914-27631187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPN1
Entrez Gene ID
11321
Gene Name
GPN-loop GTPase 1
Gene Aliases
ATPBD1A, MBDIN, NTPBP, RPAP4, XAB1
UniGene
Hs.18259
Gene Chromosome Location
Chr.2: 27628648-27650846 on Build GRCh38
Transcripts
NM_001145047.1 NM_001145048.1 NM_001145049.1 NM_007266.3 NR_026735.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6579665 c.214G>C p.D72H Chr.2: 27631035-27631035 on Build GRCh38
6046701 c.210T>C p.I70I Chr.2: 27631031-27631031 on Build GRCh38
1019845 c.211C>A p.R71S Chr.2: 27631032-27631032 on Build GRCh38
1019847 c.211C>T p.R71C Chr.2: 27631032-27631032 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771458043 Chr.2: 27631021-27631021 on Build GRCh38
rs773720302 Chr.2: 27631018-27631019 on Build GRCh38
rs183551437 Chr.2: 27631032-27631032 on Build GRCh38
rs774890414 Chr.2: 27631030-27631030 on Build GRCh38
rs776479955 Chr.2: 27631051-27631051 on Build GRCh38
rs186188470 Chr.2: 27631147-27631147 on Build GRCh38
rs776255513 Chr.2: 27631007-27631008 on Build GRCh38
rs188247609 Chr.2: 27631138-27631138 on Build GRCh38
rs770073719 Chr.2: 27631011-27631011 on Build GRCh38
rs376080375 Chr.2: 27630952-27630952 on Build GRCh38
rs563604343 Chr.2: 27631080-27631080 on Build GRCh38
rs748581559 Chr.2: 27631010-27631011 on Build GRCh38
rs749684709 Chr.2: 27631019-27631019 on Build GRCh38
rs751039060 Chr.2: 27631105-27631105 on Build GRCh38
rs992954889 Chr.2: 27631006-27631006 on Build GRCh38
rs182246150 Chr.2: 27631133-27631133 on Build GRCh38
rs980674810 Chr.2: 27630945-27630945 on Build GRCh38
rs780769999 Chr.2: 27631111-27631111 on Build GRCh38
rs761923869 Chr.2: 27630963-27630963 on Build GRCh38
rs932276607 Chr.2: 27631060-27631060 on Build GRCh38
rs548172917 Chr.2: 27631072-27631072 on Build GRCh38
rs758437689 Chr.2: 27631091-27631091 on Build GRCh38
rs368581333 Chr.2: 27631139-27631139 on Build GRCh38
rs374762641 Chr.2: 27631065-27631065 on Build GRCh38
rs764224626 Chr.2: 27631049-27631049 on Build GRCh38
rs1008995369 Chr.2: 27631113-27631113 on Build GRCh38
rs778134670 Chr.2: 27630984-27630984 on Build GRCh38
rs552440092 Chr.2: 27631136-27631136 on Build GRCh38
rs763315204 Chr.2: 27631051-27631053 on Build GRCh38
rs79898842 Chr.2: 27631109-27631109 on Build GRCh38
rs945452627 Chr.2: 27631140-27631140 on Build GRCh38
rs775567920 Chr.2: 27631033-27631033 on Build GRCh38
rs760892571 Chr.2: 27631039-27631039 on Build GRCh38
rs750441581 Chr.2: 27631093-27631093 on Build GRCh38
rs907963858 Chr.2: 27631009-27631009 on Build GRCh38
rs1021337566 Chr.2: 27630975-27630975 on Build GRCh38
rs890159028 Chr.2: 27631112-27631112 on Build GRCh38
rs368101324 Chr.2: 27631104-27631104 on Build GRCh38

View Full Product Details