Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 129434432-129434679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STRIP2
Entrez Gene ID
57464
Gene Name
striatin interacting protein 2
Gene Aliases
FAM40B, FAR11B
UniGene
Hs.489988
Gene Chromosome Location
Chr.7: 129434433-129488399 on Build GRCh38
Transcripts
NM_001134336.1 NM_020704.2 XM_006716069.3 XM_011516435.1 XM_017012470.1 XM_017012472.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6391808 c.119G>A p.R40Q Chr.7: 129434591-129434591 on Build GRCh38
6391236 c.73A>G p.K25E Chr.7: 129434545-129434545 on Build GRCh38
6391809 c.119G>A p.R40Q Chr.7: 129434591-129434591 on Build GRCh38
6391237 c.73A>G p.K25E Chr.7: 129434545-129434545 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs776956833 Chr.7: 129434595-129434595 on Build GRCh38
rs991662416 Chr.7: 129434541-129434541 on Build GRCh38
rs913071142 Chr.7: 129434603-129434603 on Build GRCh38
rs200680852 Chr.7: 129434594-129434594 on Build GRCh38
rs764449249 Chr.7: 129434649-129434649 on Build GRCh38
rs138737421 Chr.7: 129434573-129434573 on Build GRCh38
rs983011529 Chr.7: 129434478-129434478 on Build GRCh38
rs752817789 Chr.7: 129434637-129434637 on Build GRCh38
rs908403440 Chr.7: 129434508-129434508 on Build GRCh38
rs879581055 Chr.7: 129434524-129434524 on Build GRCh38
rs748826780 Chr.7: 129434463-129434463 on Build GRCh38
rs748125456 Chr.7: 129434499-129434499 on Build GRCh38
rs1002149863 Chr.7: 129434612-129434612 on Build GRCh38
rs890381240 Chr.7: 129434617-129434618 on Build GRCh38
rs773829057 Chr.7: 129434482-129434482 on Build GRCh38
rs34266807 Chr.7: 129434508-129434509 on Build GRCh38
rs949253925 Chr.7: 129434547-129434547 on Build GRCh38
rs771901284 Chr.7: 129434501-129434501 on Build GRCh38
rs112493792 Chr.7: 129434613-129434613 on Build GRCh38
rs567050501 Chr.7: 129434558-129434558 on Build GRCh38
rs931241501 Chr.7: 129434491-129434491 on Build GRCh38
rs928909729 Chr.7: 129434481-129434481 on Build GRCh38
rs986923047 Chr.7: 129434459-129434459 on Build GRCh38
rs1017875172 Chr.7: 129434644-129434644 on Build GRCh38
rs1009810326 Chr.7: 129434652-129434652 on Build GRCh38
rs919828283 Chr.7: 129434470-129434470 on Build GRCh38
rs780218934 Chr.7: 129434534-129434535 on Build GRCh38
rs975088812 Chr.7: 129434468-129434468 on Build GRCh38
rs947142317 Chr.7: 129434559-129434559 on Build GRCh38
rs759030066 Chr.7: 129434646-129434646 on Build GRCh38
rs760332304 Chr.7: 129434566-129434566 on Build GRCh38
rs759805124 Chr.7: 129434609-129434609 on Build GRCh38
rs768106111 Chr.7: 129434473-129434473 on Build GRCh38
rs938822764 Chr.7: 129434503-129434503 on Build GRCh38
rs551874048 Chr.7: 129434462-129434462 on Build GRCh38
rs1007309779 Chr.7: 129434639-129434639 on Build GRCh38
rs894060279 Chr.7: 129434643-129434643 on Build GRCh38

View Full Product Details