Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12940789-12941044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALR
Entrez Gene ID
811
Gene Name
calreticulin
Gene Aliases
CRT, HEL-S-99n, RO, SSA, cC1qR
UniGene
Hs.515162
Gene Chromosome Location
Chr.19: 12938600-12944490 on Build GRCh38
Transcripts
NM_004343.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2817018 c.909C>T p.P303P Chr.19: 12940836-12940836 on Build GRCh38
6570405 c.944G>C p.G315A Chr.19: 12940871-12940871 on Build GRCh38
6561953 c.960+3G>A p.? Chr.19: 12940890-12940890 on Build GRCh38
991546 c.899C>A p.S300Y Chr.19: 12940826-12940826 on Build GRCh38
709890 c.892G>T p.E298* Chr.19: 12940819-12940819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770320546 Chr.19: 12940817-12940817 on Build GRCh38
rs973408164 Chr.19: 12940987-12940987 on Build GRCh38
rs948134148 Chr.19: 12941008-12941008 on Build GRCh38
rs760805885 Chr.19: 12940839-12940839 on Build GRCh38
rs868355405 Chr.19: 12940999-12940999 on Build GRCh38
rs779969842 Chr.19: 12940873-12940873 on Build GRCh38
rs1026947469 Chr.19: 12940864-12940864 on Build GRCh38
rs375710146 Chr.19: 12940903-12940903 on Build GRCh38
rs764176358 Chr.19: 12940843-12940843 on Build GRCh38
rs776703195 Chr.19: 12940927-12940927 on Build GRCh38
rs753701190 Chr.19: 12940848-12940848 on Build GRCh38
rs746475957 Chr.19: 12940812-12940812 on Build GRCh38
rs775741226 Chr.19: 12940837-12940837 on Build GRCh38
rs775831137 Chr.19: 12940917-12940917 on Build GRCh38
rs987732021 Chr.19: 12940955-12940955 on Build GRCh38
rs761666518 Chr.19: 12940929-12940929 on Build GRCh38
rs116396710 Chr.19: 12940948-12940948 on Build GRCh38
rs759684254 Chr.19: 12940832-12940832 on Build GRCh38
rs527790789 Chr.19: 12940973-12940973 on Build GRCh38
rs111306057 Chr.19: 12940863-12940863 on Build GRCh38
rs199996394 Chr.19: 12940919-12940919 on Build GRCh38
rs371489238 Chr.19: 12940900-12940900 on Build GRCh38
rs749756031 Chr.19: 12940897-12940897 on Build GRCh38
rs371988722 Chr.19: 12940830-12940830 on Build GRCh38
rs920647079 Chr.19: 12940995-12940995 on Build GRCh38
rs375135016 Chr.19: 12940831-12940831 on Build GRCh38
rs964090332 Chr.19: 12940846-12940846 on Build GRCh38
rs772370252 Chr.19: 12940835-12940835 on Build GRCh38
rs749715575 Chr.19: 12940829-12940829 on Build GRCh38
rs961998301 Chr.19: 12940975-12940975 on Build GRCh38
rs761731344 Chr.19: 12940860-12940860 on Build GRCh38
rs778115432 Chr.19: 12940818-12940818 on Build GRCh38
rs908158095 Chr.19: 12940970-12940970 on Build GRCh38
rs571491760 Chr.19: 12940943-12940943 on Build GRCh38
rs779644857 Chr.19: 12940872-12940872 on Build GRCh38
rs370016483 Chr.19: 12940875-12940875 on Build GRCh38
rs372777769 Chr.19: 12940879-12940879 on Build GRCh38
rs758072446 Chr.19: 12940869-12940869 on Build GRCh38
rs765171227 Chr.19: 12940933-12940933 on Build GRCh38
rs368896588 Chr.19: 12940907-12940907 on Build GRCh38
rs778925552 Chr.19: 12940965-12940965 on Build GRCh38
rs73505370 Chr.19: 12941015-12941015 on Build GRCh38

View Full Product Details