Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 47536659-47536918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP10D
Entrez Gene ID
57205
Gene Name
ATPase phospholipid transporting 10D (putative)
Gene Aliases
ATPVD
UniGene
Hs.437241
Gene Chromosome Location
Chr.4: 47485261-47593486 on Build GRCh38
Transcripts
NM_020453.3 XM_005248119.4 XM_005248120.4 XM_011513722.2 XM_017008472.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6167540 c.1154C>G p.P385R Chr.4: 47536696-47536696 on Build GRCh38
3604160 c.1351C>T p.R451* Chr.4: 47536893-47536893 on Build GRCh38
1723473 c.1206C>T p.F402F Chr.4: 47536748-47536748 on Build GRCh38
3604159 c.1325C>T p.T442I Chr.4: 47536867-47536867 on Build GRCh38
3604158 c.1303C>G p.L435V Chr.4: 47536845-47536845 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759945258 Chr.4: 47536779-47536779 on Build GRCh38
rs768945023 Chr.4: 47536887-47536887 on Build GRCh38
rs370204271 Chr.4: 47536869-47536869 on Build GRCh38
rs746432231 Chr.4: 47536857-47536857 on Build GRCh38
rs777251060 Chr.4: 47536856-47536856 on Build GRCh38
rs139063237 Chr.4: 47536853-47536853 on Build GRCh38
rs142744698 Chr.4: 47536718-47536718 on Build GRCh38
rs375420700 Chr.4: 47536893-47536893 on Build GRCh38
rs774487967 Chr.4: 47536701-47536706 on Build GRCh38
rs779845954 Chr.4: 47536704-47536704 on Build GRCh38
rs771911187 Chr.4: 47536802-47536803 on Build GRCh38
rs754746331 Chr.4: 47536820-47536820 on Build GRCh38
rs770275319 Chr.4: 47536862-47536862 on Build GRCh38
rs766371060 Chr.4: 47536808-47536808 on Build GRCh38
rs758527471 Chr.4: 47536699-47536699 on Build GRCh38
rs772847899 Chr.4: 47536749-47536749 on Build GRCh38
rs771471852 Chr.4: 47536743-47536743 on Build GRCh38
rs764011636 Chr.4: 47536800-47536800 on Build GRCh38
rs200956632 Chr.4: 47536777-47536777 on Build GRCh38
rs898317548 Chr.4: 47536769-47536769 on Build GRCh38
rs995865598 Chr.4: 47536773-47536773 on Build GRCh38
rs770384605 Chr.4: 47536854-47536854 on Build GRCh38
rs886982488 Chr.4: 47536841-47536841 on Build GRCh38
rs779656162 Chr.4: 47536709-47536709 on Build GRCh38
rs1022160786 Chr.4: 47536826-47536826 on Build GRCh38
rs147390403 Chr.4: 47536784-47536784 on Build GRCh38
rs761536460 Chr.4: 47536707-47536708 on Build GRCh38
rs376991791 Chr.4: 47536804-47536804 on Build GRCh38
rs778507335 Chr.4: 47536735-47536735 on Build GRCh38
rs150738679 Chr.4: 47536845-47536845 on Build GRCh38
rs548816158 Chr.4: 47536868-47536868 on Build GRCh38
rs775839118 Chr.4: 47536791-47536791 on Build GRCh38
rs747410568 Chr.4: 47536754-47536754 on Build GRCh38
rs765741714 Chr.4: 47536780-47536780 on Build GRCh38
rs753004084 Chr.4: 47536698-47536698 on Build GRCh38
rs753814845 Chr.4: 47536817-47536817 on Build GRCh38
rs778738719 Chr.4: 47536821-47536821 on Build GRCh38
rs754586853 Chr.4: 47536719-47536719 on Build GRCh38
rs764289473 Chr.4: 47536707-47536707 on Build GRCh38
rs751670908 Chr.4: 47536803-47536803 on Build GRCh38
rs149876632 Chr.4: 47536894-47536894 on Build GRCh38
rs1056790780 Chr.4: 47536765-47536765 on Build GRCh38
rs747849029 Chr.4: 47536742-47536742 on Build GRCh38
rs747797887 Chr.4: 47536831-47536831 on Build GRCh38
rs371554019 Chr.4: 47536774-47536774 on Build GRCh38
rs776065711 Chr.4: 47536816-47536816 on Build GRCh38
rs777093283 Chr.4: 47536852-47536852 on Build GRCh38
rs138737356 Chr.4: 47536708-47536708 on Build GRCh38
rs200358319 Chr.4: 47536799-47536799 on Build GRCh38

View Full Product Details