Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 45424755-45425014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX5
Entrez Gene ID
240
Gene Name
arachidonate 5-lipoxygenase
Gene Aliases
5-LO, 5-LOX, 5LPG, LOG5
UniGene
Hs.89499
Gene Chromosome Location
Chr.10: 45374166-45446121 on Build GRCh38
Transcripts
NM_000698.4 NM_001256153.2 NM_001256154.2 NM_001320861.1 NM_001320862.1 XM_017016012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4818929 c.685G>A p.E229K Chr.10: 45424983-45424983 on Build GRCh38
4661209 c.666G>A p.R222R Chr.10: 45424964-45424964 on Build GRCh38
3720777 c.664C>A p.R222R Chr.10: 45424962-45424962 on Build GRCh38
1627504 c.677A>G p.H226R Chr.10: 45424975-45424975 on Build GRCh38
6598706 c.692T>G p.L231R Chr.10: 45424990-45424990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569391096 Chr.10: 45424811-45424811 on Build GRCh38
rs374722615 Chr.10: 45424944-45424944 on Build GRCh38
rs553672395 Chr.10: 45424853-45424853 on Build GRCh38
rs747310532 Chr.10: 45424919-45424919 on Build GRCh38
rs536696950 Chr.10: 45424830-45424830 on Build GRCh38
rs766190780 Chr.10: 45424907-45424907 on Build GRCh38
rs541716813 Chr.10: 45424851-45424851 on Build GRCh38
rs545601125 Chr.10: 45424897-45424897 on Build GRCh38
rs1021020813 Chr.10: 45424868-45424868 on Build GRCh38
rs977904418 Chr.10: 45424797-45424797 on Build GRCh38
rs774179264 Chr.10: 45424948-45424948 on Build GRCh38
rs759477547 Chr.10: 45424963-45424963 on Build GRCh38
rs371348748 Chr.10: 45424931-45424931 on Build GRCh38
rs77666700 Chr.10: 45424791-45424791 on Build GRCh38
rs1018867487 Chr.10: 45424813-45424813 on Build GRCh38
rs1037160589 Chr.10: 45424938-45424938 on Build GRCh38
rs769496743 Chr.10: 45424924-45424924 on Build GRCh38
rs921580135 Chr.10: 45424880-45424880 on Build GRCh38
rs374061729 Chr.10: 45424914-45424914 on Build GRCh38
rs1049707316 Chr.10: 45424783-45424783 on Build GRCh38
rs767328383 Chr.10: 45424968-45424968 on Build GRCh38
rs912802592 Chr.10: 45424902-45424902 on Build GRCh38
rs752588031 Chr.10: 45424970-45424970 on Build GRCh38
rs1010023389 Chr.10: 45424838-45424838 on Build GRCh38
rs578225550 Chr.10: 45424871-45424871 on Build GRCh38
rs868088107 Chr.10: 45424934-45424934 on Build GRCh38
rs369930922 Chr.10: 45424972-45424972 on Build GRCh38
rs573519264 Chr.10: 45424848-45424848 on Build GRCh38
rs901844916 Chr.10: 45424818-45424818 on Build GRCh38
rs367872113 Chr.10: 45424929-45424929 on Build GRCh38
rs555334569 Chr.10: 45424840-45424840 on Build GRCh38
rs987042761 Chr.10: 45424901-45424901 on Build GRCh38

View Full Product Details