Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7850056-7850287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVI5L
Entrez Gene ID
115704
Gene Name
ecotropic viral integration site 5 like
Gene Aliases
-
UniGene
Hs.26870
Gene Chromosome Location
Chr.19: 7830275-7864976 on Build GRCh38
Transcripts
NM_001159944.2 NM_145245.4 XM_005272458.4 XM_017026239.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7129040 c.715C>A p.L239I Chr.19: 7850084-7850084 on Build GRCh38
5793614 c.692T>A p.L231H Chr.19: 7850061-7850061 on Build GRCh38
6292415 c.750G>A p.L250L Chr.19: 7850119-7850119 on Build GRCh38
3668388 c.720G>A p.G240G Chr.19: 7850089-7850089 on Build GRCh38
7366718 c.712G>A p.E238K Chr.19: 7850081-7850081 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748600497 Chr.19: 7850135-7850150 on Build GRCh38
rs971826361 Chr.19: 7850249-7850249 on Build GRCh38
rs1057128082 Chr.19: 7850182-7850182 on Build GRCh38
rs981525421 Chr.19: 7850250-7850250 on Build GRCh38
rs762669875 Chr.19: 7850081-7850081 on Build GRCh38
rs375512695 Chr.19: 7850150-7850150 on Build GRCh38
rs931235647 Chr.19: 7850100-7850100 on Build GRCh38
rs904667342 Chr.19: 7850242-7850242 on Build GRCh38
rs751796861 Chr.19: 7850143-7850143 on Build GRCh38
rs1004064138 Chr.19: 7850180-7850180 on Build GRCh38
rs377173331 Chr.19: 7850135-7850135 on Build GRCh38
rs138727675 Chr.19: 7850187-7850187 on Build GRCh38
rs760686773 Chr.19: 7850119-7850119 on Build GRCh38
rs759460493 Chr.19: 7850099-7850099 on Build GRCh38
rs200789450 Chr.19: 7850134-7850134 on Build GRCh38
rs568606721 Chr.19: 7850183-7850183 on Build GRCh38
rs886924027 Chr.19: 7850174-7850174 on Build GRCh38
rs116138693 Chr.19: 7850211-7850211 on Build GRCh38
rs577083649 Chr.19: 7850234-7850234 on Build GRCh38
rs774283177 Chr.19: 7850089-7850089 on Build GRCh38
rs372190141 Chr.19: 7850080-7850080 on Build GRCh38
rs780317405 Chr.19: 7850138-7850138 on Build GRCh38
rs111420117 Chr.19: 7850169-7850169 on Build GRCh38
rs143142461 Chr.19: 7850086-7850086 on Build GRCh38
rs781641526 Chr.19: 7850167-7850167 on Build GRCh38
rs548557570 Chr.19: 7850159-7850159 on Build GRCh38
rs752538532 Chr.19: 7850111-7850111 on Build GRCh38
rs751471380 Chr.19: 7850191-7850191 on Build GRCh38
rs765209807 Chr.19: 7850130-7850130 on Build GRCh38
rs138346986 Chr.19: 7850107-7850107 on Build GRCh38
rs756859436 Chr.19: 7850134-7850135 on Build GRCh38
rs780800302 Chr.19: 7850143-7850158 on Build GRCh38
rs745348232 Chr.19: 7850156-7850157 on Build GRCh38

View Full Product Details