Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 165682052-165682309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH9A1
Entrez Gene ID
223
Gene Name
aldehyde dehydrogenase 9 family member A1
Gene Aliases
ALDH4, ALDH7, ALDH9, E3, TMABADH
UniGene
Hs.2533
Gene Chromosome Location
Chr.1: 165662212-165698663 on Build GRCh38
Transcripts
NM_000696.3 XM_011509294.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4499753 c.549C>T p.P183P Chr.1: 165682150-165682150 on Build GRCh38
5544460 c.396C>T p.I132I Chr.1: 165682231-165682231 on Build GRCh38
7299906 c.424A>G p.T142A Chr.1: 165682203-165682203 on Build GRCh38
6059250 c.422A>G p.Y141C Chr.1: 165682205-165682205 on Build GRCh38
4499754 c.477C>T p.P159P Chr.1: 165682150-165682150 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757294828 Chr.1: 165682121-165682121 on Build GRCh38
rs907151508 Chr.1: 165682093-165682093 on Build GRCh38
rs893169407 Chr.1: 165682189-165682189 on Build GRCh38
rs776795658 Chr.1: 165682255-165682255 on Build GRCh38
rs549670585 Chr.1: 165682152-165682152 on Build GRCh38
rs977121415 Chr.1: 165682220-165682220 on Build GRCh38
rs758398469 Chr.1: 165682235-165682235 on Build GRCh38
rs556696089 Chr.1: 165682262-165682262 on Build GRCh38
rs1913844 Chr.1: 165682265-165682265 on Build GRCh38
rs764369877 Chr.1: 165682194-165682194 on Build GRCh38
rs549354876 Chr.1: 165682245-165682245 on Build GRCh38
rs769013954 Chr.1: 165682137-165682137 on Build GRCh38
rs571861115 Chr.1: 165682118-165682118 on Build GRCh38
rs370991627 Chr.1: 165682282-165682282 on Build GRCh38
rs760003487 Chr.1: 165682180-165682180 on Build GRCh38
rs760770069 Chr.1: 165682214-165682214 on Build GRCh38
rs77790268 Chr.1: 165682153-165682153 on Build GRCh38
rs201328563 Chr.1: 165682127-165682127 on Build GRCh38
rs756263931 Chr.1: 165682087-165682087 on Build GRCh38
rs761164961 Chr.1: 165682191-165682191 on Build GRCh38
rs200499399 Chr.1: 165682126-165682126 on Build GRCh38
rs372810666 Chr.1: 165682114-165682114 on Build GRCh38
rs746880243 Chr.1: 165682143-165682143 on Build GRCh38
rs776283835 Chr.1: 165682149-165682149 on Build GRCh38
rs115525112 Chr.1: 165682275-165682275 on Build GRCh38
rs772742063 Chr.1: 165682154-165682154 on Build GRCh38
rs917670564 Chr.1: 165682086-165682086 on Build GRCh38
rs934953351 Chr.1: 165682144-165682144 on Build GRCh38
rs772684487 Chr.1: 165682272-165682272 on Build GRCh38
rs61757688 Chr.1: 165682151-165682151 on Build GRCh38
rs370149077 Chr.1: 165682213-165682213 on Build GRCh38
rs201921782 Chr.1: 165682280-165682280 on Build GRCh38
rs375080663 Chr.1: 165682274-165682274 on Build GRCh38
rs780207254 Chr.1: 165682241-165682241 on Build GRCh38
rs775997700 Chr.1: 165682279-165682279 on Build GRCh38
rs371946171 Chr.1: 165682142-165682142 on Build GRCh38
rs577886592 Chr.1: 165682210-165682210 on Build GRCh38
rs1021346010 Chr.1: 165682184-165682184 on Build GRCh38
rs949139643 Chr.1: 165682109-165682109 on Build GRCh38
rs542747267 Chr.1: 165682271-165682271 on Build GRCh38
rs768058910 Chr.1: 165682182-165682182 on Build GRCh38
rs775815145 Chr.1: 165682183-165682183 on Build GRCh38
rs543683476 Chr.1: 165682249-165682249 on Build GRCh38

View Full Product Details