Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 22256575-22256824 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEF2K
Entrez Gene ID
29904
Gene Name
eukaryotic elongation factor 2 kinase
Gene Aliases
HSU93850, eEF-2K
UniGene
Hs.498892
Gene Chromosome Location
Chr.16: 22206264-22288745 on Build GRCh38
Transcripts
NM_013302.3 XM_017023193.1 XM_017023194.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4431636 c.662C>A p.P221Q Chr.16: 22256791-22256791 on Build GRCh38
6078863 c.619G>T p.V207L Chr.16: 22256748-22256748 on Build GRCh38
6440917 c.630G>A p.M210I Chr.16: 22256759-22256759 on Build GRCh38
3507636 c.645C>T p.I215I Chr.16: 22256774-22256774 on Build GRCh38
5342722 c.693C>T p.Y231Y Chr.16: 22256822-22256822 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764479203 Chr.16: 22256747-22256747 on Build GRCh38
rs889524237 Chr.16: 22256649-22256649 on Build GRCh38
rs773029107 Chr.16: 22256800-22256800 on Build GRCh38
rs766307698 Chr.16: 22256740-22256740 on Build GRCh38
rs138681546 Chr.16: 22256791-22256791 on Build GRCh38
rs956899933 Chr.16: 22256605-22256605 on Build GRCh38
rs749730972 Chr.16: 22256704-22256704 on Build GRCh38
rs780591039 Chr.16: 22256774-22256774 on Build GRCh38
rs778036162 Chr.16: 22256700-22256700 on Build GRCh38
rs970100511 Chr.16: 22256767-22256767 on Build GRCh38
rs952153699 Chr.16: 22256721-22256721 on Build GRCh38
rs779375595 Chr.16: 22256762-22256762 on Build GRCh38
rs1030388373 Chr.16: 22256662-22256662 on Build GRCh38
rs761596131 Chr.16: 22256730-22256730 on Build GRCh38
rs755211834 Chr.16: 22256689-22256689 on Build GRCh38
rs754046649 Chr.16: 22256755-22256755 on Build GRCh38
rs764950680 Chr.16: 22256732-22256732 on Build GRCh38
rs559638756 Chr.16: 22256687-22256687 on Build GRCh38
rs1031016648 Chr.16: 22256602-22256602 on Build GRCh38
rs376768453 Chr.16: 22256741-22256741 on Build GRCh38
rs762785586 Chr.16: 22256803-22256803 on Build GRCh38
rs909152441 Chr.16: 22256696-22256696 on Build GRCh38
rs369121898 Chr.16: 22256773-22256773 on Build GRCh38
rs938311264 Chr.16: 22256729-22256729 on Build GRCh38
rs531042317 Chr.16: 22256765-22256765 on Build GRCh38
rs771638359 Chr.16: 22256709-22256709 on Build GRCh38
rs762759374 Chr.16: 22256738-22256738 on Build GRCh38
rs757711107 Chr.16: 22256760-22256760 on Build GRCh38
rs750681800 Chr.16: 22256740-22256741 on Build GRCh38
rs961325000 Chr.16: 22256701-22256701 on Build GRCh38
rs372767330 Chr.16: 22256706-22256706 on Build GRCh38
rs773153979 Chr.16: 22256735-22256735 on Build GRCh38
rs776259440 Chr.16: 22256724-22256724 on Build GRCh38
rs368760106 Chr.16: 22256705-22256705 on Build GRCh38
rs112106407 Chr.16: 22256792-22256792 on Build GRCh38
rs202192956 Chr.16: 22256720-22256720 on Build GRCh38
rs375099811 Chr.16: 22256707-22256707 on Build GRCh38
rs147220043 Chr.16: 22256769-22256769 on Build GRCh38
rs180704155 Chr.16: 22256713-22256713 on Build GRCh38
rs751953174 Chr.16: 22256624-22256624 on Build GRCh38
rs746500121 Chr.16: 22256714-22256714 on Build GRCh38
rs576993932 Chr.16: 22256608-22256608 on Build GRCh38
rs985959649 Chr.16: 22256688-22256688 on Build GRCh38
rs147622817 Chr.16: 22256775-22256775 on Build GRCh38
rs545761834 Chr.16: 22256675-22256675 on Build GRCh38
rs751003245 Chr.16: 22256763-22256763 on Build GRCh38
rs1028510805 Chr.16: 22256621-22256621 on Build GRCh38

View Full Product Details