Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123980586-123980843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FER1L6
Entrez Gene ID
654463
Gene Name
fer-1 like family member 6
Gene Aliases
C8ORFK23
UniGene
Hs.632058
Gene Chromosome Location
Chr.8: 123851987-124120061 on Build GRCh38
Transcripts
NM_001039112.2 XM_006716618.3 XM_011517231.2 XM_011517232.2 XM_011517233.2 XM_011517235.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3644878 c.1277C>T p.S426F Chr.8: 123980678-123980678 on Build GRCh38
3644879 c.1282G>A p.D428N Chr.8: 123980683-123980683 on Build GRCh38
317360 c.1368C>A p.N456K Chr.8: 123980769-123980769 on Build GRCh38
4769793 c.1226G>A p.R409Q Chr.8: 123980627-123980627 on Build GRCh38
3644880 c.1337G>A p.G446E Chr.8: 123980738-123980738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778956217 Chr.8: 123980749-123980749 on Build GRCh38
rs1026391512 Chr.8: 123980801-123980801 on Build GRCh38
rs372652726 Chr.8: 123980807-123980807 on Build GRCh38
rs113087741 Chr.8: 123980793-123980793 on Build GRCh38
rs950533888 Chr.8: 123980801-123980802 on Build GRCh38
rs78034602 Chr.8: 123980791-123980791 on Build GRCh38
rs374214825 Chr.8: 123980681-123980681 on Build GRCh38
rs760684546 Chr.8: 123980655-123980655 on Build GRCh38
rs765650846 Chr.8: 123980805-123980805 on Build GRCh38
rs758857875 Chr.8: 123980686-123980686 on Build GRCh38
rs755625470 Chr.8: 123980701-123980701 on Build GRCh38
rs759710529 Chr.8: 123980804-123980804 on Build GRCh38
rs934724992 Chr.8: 123980627-123980627 on Build GRCh38
rs765815577 Chr.8: 123980670-123980670 on Build GRCh38
rs768448259 Chr.8: 123980612-123980612 on Build GRCh38
rs576479024 Chr.8: 123980656-123980656 on Build GRCh38
rs749208767 Chr.8: 123980735-123980735 on Build GRCh38
rs199707703 Chr.8: 123980744-123980744 on Build GRCh38
rs201254129 Chr.8: 123980626-123980626 on Build GRCh38
rs200225461 Chr.8: 123980709-123980709 on Build GRCh38
rs776542917 Chr.8: 123980798-123980798 on Build GRCh38
rs759995131 Chr.8: 123980665-123980665 on Build GRCh38
rs368417039 Chr.8: 123980778-123980778 on Build GRCh38
rs750103113 Chr.8: 123980699-123980699 on Build GRCh38
rs527654182 Chr.8: 123980775-123980775 on Build GRCh38
rs933328316 Chr.8: 123980727-123980727 on Build GRCh38
rs923283844 Chr.8: 123980688-123980688 on Build GRCh38
rs773368685 Chr.8: 123980792-123980792 on Build GRCh38
rs1010335853 Chr.8: 123980810-123980810 on Build GRCh38
rs754296474 Chr.8: 123980659-123980659 on Build GRCh38
rs868607365 Chr.8: 123980632-123980632 on Build GRCh38
rs201686117 Chr.8: 123980647-123980647 on Build GRCh38
rs1057028986 Chr.8: 123980630-123980630 on Build GRCh38
rs775161966 Chr.8: 123980723-123980723 on Build GRCh38
rs780988475 Chr.8: 123980696-123980696 on Build GRCh38
rs761706612 Chr.8: 123980629-123980629 on Build GRCh38
rs772158690 Chr.8: 123980638-123980638 on Build GRCh38
rs770962496 Chr.8: 123980797-123980797 on Build GRCh38
rs750047724 Chr.8: 123980808-123980808 on Build GRCh38

View Full Product Details