Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 72350457-72350728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEXA
Entrez Gene ID
3073
Gene Name
hexosaminidase subunit alpha
Gene Aliases
TSD
UniGene
Hs.604479
Gene Chromosome Location
Chr.15: 72343435-72376473 on Build GRCh38
Transcripts
NM_000520.5 NM_001318825.1 NR_134869.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3744738 c.704C>T p.T235I Chr.15: 72350619-72350619 on Build GRCh38
5591258 c.693C>T p.T231T Chr.15: 72350630-72350630 on Build GRCh38
6464056 c.759G>A p.V253V Chr.15: 72350564-72350564 on Build GRCh38
4507688 c.753C>T p.I251I Chr.15: 72350570-72350570 on Build GRCh38
4690170 c.746G>A p.R249Q Chr.15: 72350577-72350577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774544364 Chr.15: 72350692-72350692 on Build GRCh38
rs77405894 Chr.15: 72350576-72350576 on Build GRCh38
rs780189623 Chr.15: 72350659-72350659 on Build GRCh38
rs757119445 Chr.15: 72350617-72350617 on Build GRCh38
rs75211071 Chr.15: 72350663-72350663 on Build GRCh38
rs121907980 Chr.15: 72350517-72350517 on Build GRCh38
rs769866128 Chr.15: 72350646-72350646 on Build GRCh38
rs121907954 Chr.15: 72350518-72350518 on Build GRCh38
rs533004263 Chr.15: 72350560-72350560 on Build GRCh38
rs762299363 Chr.15: 72350699-72350699 on Build GRCh38
rs746942625 Chr.15: 72350528-72350528 on Build GRCh38
rs121907970 Chr.15: 72350584-72350584 on Build GRCh38
rs370576428 Chr.15: 72350559-72350559 on Build GRCh38
rs34093438 Chr.15: 72350670-72350671 on Build GRCh38
rs867517363 Chr.15: 72350498-72350498 on Build GRCh38
rs1057519462 Chr.15: 72350535-72350535 on Build GRCh38
rs771004345 Chr.15: 72350532-72350532 on Build GRCh38
rs774868739 Chr.15: 72350566-72350566 on Build GRCh38
rs566580738 Chr.15: 72350569-72350569 on Build GRCh38
rs745941151 Chr.15: 72350631-72350631 on Build GRCh38
rs567614636 Chr.15: 72350695-72350695 on Build GRCh38
rs747291156 Chr.15: 72350604-72350604 on Build GRCh38
rs757023807 Chr.15: 72350661-72350661 on Build GRCh38
rs138058578 Chr.15: 72350578-72350578 on Build GRCh38
rs778770571 Chr.15: 72350497-72350497 on Build GRCh38
rs117513345 Chr.15: 72350564-72350564 on Build GRCh38
rs762255098 Chr.15: 72350568-72350568 on Build GRCh38
rs371038167 Chr.15: 72350686-72350686 on Build GRCh38
rs758166013 Chr.15: 72350587-72350587 on Build GRCh38
rs150675340 Chr.15: 72350614-72350614 on Build GRCh38
rs776660720 Chr.15: 72350547-72350547 on Build GRCh38
rs747954846 Chr.15: 72350509-72350509 on Build GRCh38
rs769020800 Chr.15: 72350676-72350676 on Build GRCh38
rs755203085 Chr.15: 72350699-72350699 on Build GRCh38
rs758135123 Chr.15: 72350511-72350511 on Build GRCh38
rs916204955 Chr.15: 72350541-72350541 on Build GRCh38
rs121907971 Chr.15: 72350551-72350551 on Build GRCh38
rs746861846 Chr.15: 72350611-72350611 on Build GRCh38
rs991742498 Chr.15: 72350520-72350520 on Build GRCh38
rs121907959 Chr.15: 72350574-72350574 on Build GRCh38
rs76873846 Chr.15: 72350582-72350582 on Build GRCh38
rs777460475 Chr.15: 72350588-72350588 on Build GRCh38
rs567489072 Chr.15: 72350701-72350701 on Build GRCh38
rs747237410 Chr.15: 72350681-72350681 on Build GRCh38
rs745997044 Chr.15: 72350548-72350548 on Build GRCh38
rs587779407 Chr.15: 72350604-72350605 on Build GRCh38
rs781164103 Chr.15: 72350618-72350618 on Build GRCh38
rs571944965 Chr.15: 72350490-72350490 on Build GRCh38

View Full Product Details