Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 37325875-37326100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NWD2
Entrez Gene ID
57495
Gene Name
NACHT and WD repeat domain containing 2
Gene Aliases
KIAA1239
UniGene
Hs.4280
Gene Chromosome Location
Chr.4: 37245068-37449465 on Build GRCh38
Transcripts
NM_001144990.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4536251 c.227G>C p.G76A Chr.4: 37326011-37326011 on Build GRCh38
2845948 c.155C>T p.T52M Chr.4: 37325939-37325939 on Build GRCh38
7373615 c.190T>C p.Y64H Chr.4: 37325974-37325974 on Build GRCh38
5897497 c.157G>A p.G53R Chr.4: 37325941-37325941 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs911610279 Chr.4: 37326055-37326055 on Build GRCh38
rs771873209 Chr.4: 37325972-37325972 on Build GRCh38
rs73130327 Chr.4: 37325961-37325961 on Build GRCh38
rs749236042 Chr.4: 37325991-37325991 on Build GRCh38
rs368673772 Chr.4: 37325940-37325940 on Build GRCh38
rs776716770 Chr.4: 37325979-37325979 on Build GRCh38
rs879131596 Chr.4: 37325987-37325987 on Build GRCh38
rs766188257 Chr.4: 37325978-37325978 on Build GRCh38
rs780113429 Chr.4: 37325955-37325955 on Build GRCh38
rs200417524 Chr.4: 37326034-37326034 on Build GRCh38
rs1035983855 Chr.4: 37326048-37326048 on Build GRCh38
rs772933993 Chr.4: 37325973-37325973 on Build GRCh38
rs745877866 Chr.4: 37325939-37325939 on Build GRCh38
rs764905992 Chr.4: 37326037-37326037 on Build GRCh38
rs539754619 Chr.4: 37325957-37325957 on Build GRCh38
rs781736216 Chr.4: 37325912-37325912 on Build GRCh38
rs769801939 Chr.4: 37325913-37325913 on Build GRCh38
rs888501727 Chr.4: 37325921-37325921 on Build GRCh38
rs1014479246 Chr.4: 37325933-37325933 on Build GRCh38
rs1037563874 Chr.4: 37325958-37325958 on Build GRCh38
rs752364503 Chr.4: 37326044-37326044 on Build GRCh38
rs985924673 Chr.4: 37326042-37326042 on Build GRCh38

View Full Product Details