Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 74738487-74738712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFM2
Entrez Gene ID
84340
Gene Name
G elongation factor mitochondrial 2
Gene Aliases
EF-G2mt, EFG2, MRRF2, MST027, MSTP027, RRF2, RRF2mt, hEFG2, mEF-G 2
UniGene
Hs.277154
Gene Chromosome Location
Chr.5: 74721204-74767371 on Build GRCh38
Transcripts
NM_001281302.1 NM_032380.4 NM_170681.2 NM_170691.2 NR_104006.1 XM_011543691.2 XM_017009986.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3856278 c.1159C>T p.R387C Chr.5: 74738563-74738563 on Build GRCh38
5174522 c.1155delT p.F385fs*9 Chr.5: 74738567-74738567 on Build GRCh38
6353673 c.1196T>C p.I399T Chr.5: 74738526-74738526 on Build GRCh38
3073216 c.1127A>G p.H376R Chr.5: 74738595-74738595 on Build GRCh38
5652030 c.1213A>C p.N405H Chr.5: 74738509-74738509 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147925896 Chr.5: 74738534-74738534 on Build GRCh38
rs778605094 Chr.5: 74738683-74738683 on Build GRCh38
rs763038574 Chr.5: 74738523-74738523 on Build GRCh38
rs561517778 Chr.5: 74738680-74738680 on Build GRCh38
rs752142132 Chr.5: 74738684-74738684 on Build GRCh38
rs191007875 Chr.5: 74738602-74738602 on Build GRCh38
rs778863487 Chr.5: 74738587-74738587 on Build GRCh38
rs747322521 Chr.5: 74738592-74738592 on Build GRCh38
rs928364160 Chr.5: 74738669-74738669 on Build GRCh38
rs752948580 Chr.5: 74738514-74738514 on Build GRCh38
rs561167614 Chr.5: 74738670-74738670 on Build GRCh38
rs759997637 Chr.5: 74738611-74738611 on Build GRCh38
rs769887272 Chr.5: 74738619-74738619 on Build GRCh38
rs374630200 Chr.5: 74738559-74738559 on Build GRCh38
rs866401948 Chr.5: 74738563-74738563 on Build GRCh38
rs753758523 Chr.5: 74738557-74738557 on Build GRCh38
rs780130081 Chr.5: 74738535-74738535 on Build GRCh38
rs112155465 Chr.5: 74738634-74738634 on Build GRCh38
rs538761639 Chr.5: 74738663-74738663 on Build GRCh38
rs118133558 Chr.5: 74738628-74738628 on Build GRCh38
rs1029102576 Chr.5: 74738662-74738662 on Build GRCh38
rs749758056 Chr.5: 74738614-74738614 on Build GRCh38
rs762693230 Chr.5: 74738661-74738661 on Build GRCh38
rs747881218 Chr.5: 74738583-74738583 on Build GRCh38
rs771592465 Chr.5: 74738584-74738584 on Build GRCh38
rs754563414 Chr.5: 74738565-74738565 on Build GRCh38
rs866244233 Chr.5: 74738610-74738610 on Build GRCh38
rs143233751 Chr.5: 74738526-74738526 on Build GRCh38
rs777129105 Chr.5: 74738608-74738608 on Build GRCh38
rs760387518 Chr.5: 74738666-74738666 on Build GRCh38
rs182115360 Chr.5: 74738667-74738667 on Build GRCh38
rs531729405 Chr.5: 74738671-74738671 on Build GRCh38
rs199681606 Chr.5: 74738575-74738575 on Build GRCh38
rs141886148 Chr.5: 74738620-74738620 on Build GRCh38

View Full Product Details